Skip to Content
Merck
All Photos(3)

Documents

SAB4200857

Sigma-Aldrich

IgM, Kappa from murine myeloma

clone TEPC 183-SY, purified from hybridoma cell culture

Synonym(s):

Murine IgM Kappa, Myeloma IgM Kappa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

TEPC 183-SY

form

liquid

concentration

2-2.2 mg/mL

technique(s)

ELISA: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

IgM antibodies are present as pentamers in the serum and are produced in response to antigens.IgM is a highly conserved antibody in vertebrates and is expressed early during immune response.

Specificity

IgM, Kappa immunoglobulin is (mouse IgM isotype) is derived from the TEPC 183-SY hybridoma, produced by the fusion of mouse NS1 myeloma and TEPC 183 mouse tumor line. TEPC 183 tumor line that produces mouse IgM, Kappa, is pristane-induced plasmacytoma.

Application

Identity and purity of the immunoglobulin is established by HPLC.

Biochem/physiol Actions

IgM plays a key role in the engulfing of apoptotic cells. A reduction in serum IgM levels leads to increased autoimmune response and higher risk for infections. IgM displays polyreactive and autoreactive functionality. It plays a key role in tissue homeostasis by mediating clearance of tissue based molecules. IgM has sites for N-linked glycosylation, with sugars namely, mannose, galactose, N-acetyl glucosamine and sialic acid.

Physical form

Supplied as a solution in 0.05M Tris, 0.5M NaCl, pH 8.0, containing 0.02% sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service