Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502010

Sigma-Aldrich

Anti-NNOS antibody produced in rabbit

affinity isolated antibody

Synonym(s):

BNOS, Constitutive NOS, N-NOS, NC-NOS, bNOS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 160 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS1(4842)

General description

Anti-NNOS antibody detects endogenous levels of total NNOS protein.
The NNOS (nitric oxide synthase 1) gene is mapped to human chromosome 12q24.22. Neuronal nitric oxide synthase is known to be expressed in skeletal muscles, the heart and the brain.

Immunogen

The antiserum was produced against synthesized peptide derived from human nNOS.

Immunogen Range: 818-867

Application

Anti-NNOS antibody produced in rabbit has been used in immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

NNOS (nitric oxide synthase 1) is responsible for the synthesis of NO (nitric oxide) as well as superoxide. Stress induces the activity of NNOS and generation of NO and results in the formation of nitrogen radicals. Elevation of nitrogen radical leads to intracellular protein damage and also induces impairment to mitochondrial transport chain components. This generally results in cellular energy deficiency. NNOS is known to regulate vasodilation in the forearm, in response to stress. Neuronal nitric oxide synthase normally functions to control the basal coronary blood flow.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideshi Ihara et al.
The Biochemical journal, 474(7), 1149-1162 (2017-01-28)
We previously demonstrated different spacial expression profiles of the neuronal nitric oxide (NO) synthase (nNOS) splice variants nNOS-µ and nNOS-α in the brain; however, their exact functions are not fully understood. Here, we used electron paramagnetic resonance to compare the
A Kumar et al.
Vitamins and hormones, 103, 147-167 (2017-01-08)
Stress is often marked by a state of hyperarousal to aid the initiation of necessary stress response for the successful management of stressful stimuli. It can be manifested as a challenge (stimulus) that requires behavioral, psychological, and physiological adaptations for
G Catanzaro et al.
Journal of neuroimmunology, 294, 32-40 (2016-05-04)
The development of multiple sclerosis, a major neurodegenerative disease, is due to both genetic and environmental factors that might trigger aberrant epigenetic changes of the genome. In this study, we analysed global DNA methylation in the brain of mice upon
Yan Yang et al.
Molecular medicine reports, 13(2), 1220-1226 (2015-12-10)
Spinal nitric oxide is involved in the mechanisms of pain generation and transmission during inflammatory and neuropathic pain. The aim of the present study was to explore the role of spinal nitric oxide in the development of bone cancer pain.
Sitara G Khan et al.
American journal of physiology. Heart and circulatory physiology, 313(3), H578-H583 (2017-06-25)
Mental stress-induced ischemia approximately doubles the risk of cardiac events in patients with coronary artery disease, yet the mechanisms underlying changes in coronary blood flow in response to mental stress are poorly characterized. Neuronal nitric oxide synthase (nNOS) regulates basal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service