Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200559

Sigma-Aldrich

Anti-VAChT (C-terminal) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-SLC18A3, Anti-Solute carrier family 18 member 3, Anti-VAT, Anti-vesicular acetylcholine transporter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect immunofluorescence: 1:1,000-1:2,000 using PC12 cells treated with rat NGF.
western blot: 1:500-1:800 using extracts of mouse brain (S1 fraction).

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Vesicular Acetylcholine Transporter (VAChT, SLC18A3) belongs to the solute carrier family 18 (SLC18) family of proton/neurotransmitter antiporters and shares a high degree of homology to vesicular monoamine transporters (VMATs). It is a 12-transmembrane domain protein. VAChT is expressed in all major cholinergic neurons in the central and peripheral nervous system and is mainly localized in small synaptic vesicles in cholinergic nerve terminals.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminus of human vesicular acetylcholine transporter (VAChT), conjugated to KLH. The corresponding sequence is highly conserved in rat and mouse VAChT (82% identity).

Application

Anti-VAChT (C-terminal) antibody produced in rabbit has been used in:
  • immunoblotting
  • immunofluorescence staining
  • immunocytochemistry
  • double immunolabeling

Biochem/physiol Actions

Vesicular Acetylcholine Transporter (VAChT), is required in cholinergic neurons for the selective transport of acetylcholine (ACh) into synaptic vesicles. It depends on the activity of a vacuolar-type H+ -ATPase to accumulate ACh in synaptic vesicles. Knockdown of VAChT in the hippocampus has been shown to cause behavior changes, learning deficits, specific deficits in spatial memory and long-term potentiation (LTP). VAChT provides a specific marker for cholinergic neurons, for the study of cholinergic transmission in experimental models of Alzheimer′s disease and other nervous system disorders. Cloning of the VAChT gene has revealed a single genetic locus encoding both VAChT and ChAT, providing a unique genomic arrangement suggesting that these genes may be co-regulated.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Li et al.
Frontiers in aging neuroscience, 13, 770948-770948 (2022-02-22)
To explore the effect of electro-acupuncture (EA) treatment on pattern separation and investigate the neural circuit mechanism involved in five familial mutations (5 × FAD) mice. Five familial mutations mice were treated with EA at Baihui (DU20) and Shenting (DU24)
Vesicular acetylcholine transporter defect underlies devastating congenital myasthenia syndrome
Aran A, et al.
Neurology, 88(11), 1021-1028 (2017)
Identification and distribution of neuronal nitric oxide synthase and neurochemical markers in the neuroepithelial cells of the gill and the skin in the giant mudskipper, Periophthalmodon schlosseri
Zaccone G, et al.
Zoology (Jena, Germany), 125, 41-52 (2017)
The vesicular acetylcholine transporter is present in melanocytes and keratinocytes in the human epidermis
Elwary SMA, et al.
The Journal of Investigative Dermatology, 126(8), 1879-1884 (2006)
Alzheimer's disease: targeting the cholinergic system
Ferreira-Vieira TH, et al.
Current Neuropharmacology, 14(1), 101-115 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service