Skip to Content
Merck
All Photos(12)

Key Documents

SAB2702132

Sigma-Aldrich

Anti-HIF1A antibody produced in rabbit

Synonym(s):

HIF1-ALPHA, MOP1, PASD8, bHLHe78

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

93 kDa

species reactivity

mouse, bovine, human, rat

concentration

1 mg/mL

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: 1:100-1:1000
immunoprecipitation (IP): suitable
western blot: 500-10000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HIF1A(3091)

Immunogen

Recombinant fragment contain a sequence corresponding to a region within amino acids 657 and 826 of HIF1 alpha

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-Fr: Assay-dependent dilution, IHC-P: 1:100-1:1000, IP: 1:100-1:570, WB: 1:500-1:10000, ChIP assay: Assay-dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Hypoxia-inducible factor-1 (HIF1) is a transcription factor found in mammalian cells cultured under reduced oxygen tension that plays an essential role in cellular and systemic homeostatic responses to hypoxia. HIF1 is a heterodimer composed of an alpha subunit and a beta subunit. The beta subunit has been identified as the aryl hydrocarbon receptor nuclear translocator (ARNT). This gene encodes the alpha subunit of HIF-1. Overexpression of a natural antisense transcript (aHIF) of this gene has been shown to be associated with nonpapillary renal carcinomas. Two alternative transcripts encoding different isoforms have been identified. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lehao Ren et al.
Virologica Sinica, 36(6), 1532-1542 (2021-09-15)
Viruses depend on host cellular metabolism to provide the energy and biosynthetic building blocks required for their replication. In this study, we observed that influenza A virus (H1N1), a single-stranded, negative-sense RNA virus with an eight-segmented genome, enhanced glycolysis both
Magdalena H Menhofer et al.
Cardiovascular research, 104(2), 303-314 (2014-09-23)
Inhibiting angiogenesis is a major approach in tumour therapy. To combat angiogenesis, the tubulin cytoskeleton has emerged as an interesting target in many pre- and clinical studies. Contrarily, the actin cytoskeleton has been largely neglected as a potential drug target
Ang Luo et al.
NAR cancer, 2(2), zcaa010-zcaa010 (2020-08-04)
Mechanistic understanding of hypoxia-responsive signaling pathways provides important insights into oxygen- and metabolism-dependent cellular phenotypes in diseases. Using SILAC-based quantitative proteomics, we provided a quantitative map identifying over 6300 protein groups in response to hypoxia in prostate cancer cells and
Lei Yang et al.
Molecular therapy. Nucleic acids, 20, 292-307 (2020-03-22)
Recently, the identification of several circular RNAs (circRNAs) as vital regulators of microRNAs (miRNAs) underlines the increasing complexity of non-coding RNA (ncRNA)-mediated regulatory networks. This study aimed to explore the effects of mmu_circ_0000790 on the biological behaviors of pulmonary artery
J S M Cuffe et al.
The Journal of physiology, 592(14), 3127-3141 (2014-05-08)
Maternal hypoxia is a common perturbation that can disrupt placental and thus fetal development, contributing to neonatal impairments. Recently, evidence has suggested that physiological outcomes are dependent upon the sex of the fetus, with males more susceptible to hypoxic insults

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service