Skip to Content
Merck
All Photos(6)

Key Documents

HPA020028

Sigma-Aldrich

Anti-RYR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cardiac muscle ryanodine receptor-calcium release channel, Anti-Cardiac muscle-type ryanodine receptor, Anti-RYR-2, Anti-RyR2, Anti-Ryanodine receptor 2, Anti-hRYR-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1,000

immunogen sequence

QDEVRGDGEEGERKPLEAALPSEDLTDLKELTEESDLLSDIFGLDLKREGGQYKLIPHNPNAGLSDLMSNPVPMPEVQEKFQEQKAKEEEKEEKEETKSEPE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RYR2(6262)

General description

RYR2 (ryanodine receptor 2) is an intracellular Ca2+ release channel present on the sarcoplasmic reticulum (SR).
RYR2 gene is located at the human chromosome location 1q43. It is majorly expressed in the brain and heart.

Immunogen

Ryanodine receptor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RYR2 (ryanodine receptor 2) provides guidance to the release and transport of Ca2+ from sarcoplasmic reticulum (SR) to the cytoplasm during cardiac muscle excitation-contraction (EC) coupling. Protein kinase A (PKA) phosphorylated RYR2 separates (FKBP12.6) to regulate the channel open probability (Po). During the process, a small portion of Ca2+ enter into the cell through the L-type Ca2+ channel, which further activates the RyR2 channel upon membrane depolarization. The activated RyR2 channel releases a large amount of Ca2+ from the SR and subsequent muscle contraction. Missense mutations in this gene cause Kazakh idiopathic ventricular tachycardia and arrhythmogenic right ventricular dysplasia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73317

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siew Mei Yap et al.
Seizure, 67, 11-14 (2019-03-09)
Ryanodine receptor 2 (RYR2) mutation is well-established in the aetiology of an inherited cardiac disorder known as catecholaminergic polymorphic ventricular tachycardia (CPVT). The RYR2 receptor is expressed in cardiomyocytes, and also in the hippocampus. The RYR2 mutation has not been
Structural basis for the gating mechanism of the type 2 ryanodine receptor RyR2
Peng W, et al.
Science, 354(6310), aah5324-aah5324 (2016)
Miriam E Hurley et al.
Methods (San Diego, Calif.), 193, 27-37 (2020-10-16)
Nanometre-scale cellular information obtained through super-resolution microscopies are often unaccompanied by functional information, particularly transient and diffusible signals through which life is orchestrated in the nano-micrometre spatial scale. We describe a correlative imaging protocol which allows the ubiquitous intracellular second
Claire Poulet et al.
Cardiovascular research (2020-02-14)
In cardiomyocytes, transverse tubules (T-tubules) associate with the sarcoplasmic reticulum (SR), forming junctional membrane complexes (JMCs) where L-type calcium channels (LTCCs) are juxtaposed to Ryanodine receptors (RyR). Junctophilin-2 (JPH2) supports the assembly of JMCs by tethering T-tubules to the SR
Michelle L Munro et al.
Frontiers in cell and developmental biology, 9, 633704-633704 (2021-03-16)
The release of Ca2+ by ryanodine receptor (RyR2) channels is critical for cardiac function. However, abnormal RyR2 activity has been linked to the development of arrhythmias, including increased spontaneous Ca2+ release in human atrial fibrillation (AF). Clustering properties of RyR2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service