Skip to Content
Merck
All Photos(4)

Documents

SAB2501100

Sigma-Aldrich

Anti-Vitamin D-Binding Protein/GC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DBP, Anti-DBP/GC, Anti-VDBG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GC(2638)

Immunogen

Peptide with sequence CDNLSTKNSKFED from the internal region of the protein sequence according to NP_000574.2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralf Herwig et al.
Biomedicines, 10(8) (2022-07-28)
Vitamin D3 complexed to deglycosylated vitamin D binding protein (VitD-dgVDBP) is a water-soluble vitamin D dimeric compound (VitD-dgVDBP). It is not clear how VitD-dgVDBP affects circulating monocytes, macrophages, other immune cell systems, including phagocytosis and apoptosis, and the generation of
Martin Kongsbak et al.
BMC immunology, 15, 35-35 (2014-09-19)
In vitro studies have shown that the active form of vitamin D3, 1α,25-dihydroxyvitamin D3 (1,25(OH)2D3), can regulate differentiation of CD4+ T cells by inhibiting Th1 and Th17 cell differentiation and promoting Th2 and Treg cell differentiation. However, the serum concentration
Simona Perga et al.
PloS one, 10(6), e0129291-e0129291 (2015-06-06)
Multiple sclerosis (MS) is a multifactorial autoimmune disease of the central nervous system with a heterogeneous and unpredictable course. To date there are no prognostic biomarkers even if they would be extremely useful for early patient intervention with personalized therapies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service