Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503071

Sigma-Aldrich

Anti-Vitamin D Receptor antibody produced in rabbit

affinity isolated antibody

Synonym(s):

1,25-dihydroxyvitamin D3 receptor, NR1I1, vitamin D receptor, vitamin D3 receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VDR(7421)

General description

Anti-Vitamin D Receptor Antibody detects endogenous levels of total Vitamin D Receptor protein.
Vitamin D receptor (VDR) gene codes for a nuclear receptor. It is expressed at a high level in the hypothalamus and the large neurons of the substantia nigra in the brain. VDR gene comprises 14 exons and 6 untranslated exons with alternative splicing sites. It is mapped to human chromosome 12q13.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Vitamin D Receptor.

Immunogen Range: 181-230

Application

Anti-Vitamin D Receptor antibody produced in rabbit has been used in western blotting and immunofluorescence staining (1:200) (1:100).

Biochem/physiol Actions

Vitamin D receptor (VDR) protein modulates genes associated with inflammation, chronic diseases, oxidative damage, and the metabolism of bone. VDR gene is involved in the susceptibility of type 2 diabetes mellitus (T2DM). The nuclear receptor encoded by the VDR gene possesses pluripotent effects. It is capable of specifically binding the elements of DNA response when bound to the ligand. VDR acts as a receptor for 1,25 dihydroxyvitamin D3 (1,25(OH)2 D3). It facilitates the biological activities of vitamin D. Inadequacy of vitamin D is a risk factor for Alzheimer′s disease (AD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Del Puerto et al.
The British journal of dermatology, 179(1), 95-100 (2017-11-07)
Vitamin D deficiency is associated with higher risk of cancer, possibly due to its antiproliferative, antiangiogenic, proapoptotic, cell-differentiating and anti-invasive effects. The anticarcinogenic role of vitamin D in melanoma is still a matter of debate. Loss of nuclear and cytoplasmic
Charlotte Gröschel et al.
Nutrients, 12(1) (2019-12-28)
Inflammatory bowel disease increases the odds of developing colitis-associated cancer. We hypothesized that Western-style diet (WD) aggravates azoxymethane (AOM)/dextran sulfate sodium salt (DSS)-induced colitis-associated tumorigenesis and that switching to the standard AIN93G diet will ameliorate disease symptoms even after cancer
Julia Höbaus et al.
International journal of cancer, 138(2), 440-450 (2015-08-05)
Our previous studies showed that the 1,25-dihydroxyvitamin D (1,25-D3) catabolizing enzyme, 1,25-dihydoxyvitamin D 24 hydroxylase (CYP24A1) was overexpressed in colorectal tumours and its level correlated with increased proliferation. We hypothesised that cells overexpressing CYP24A1 have growth advantage and a diet
Hamid Reza Khorram Khorshid et al.
Iranian journal of public health, 42(11), 1253-1258 (2013-11-01)
Late-onset Alzheimer's disease (AD), a genetically heterogeneous neurodegenerative disorder, is the most common form of dementia in people over 65 years old. The role of vitamin D in neuropsychiatric and neurodegenerative disorders such as AD has been supported by epidemiologic
Julie De Smedt et al.
Melanoma research, 34(2), 125-133 (2024-02-13)
Decrease of vitamin D receptor (VDR) expression is observed in melanocytic naevi and melanoma compared to normal skin. Little is known about factors influencing VDR expression in cutaneous melanoma (CM). We investigated the correlation of VDR expression in CM with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service