Skip to Content
Merck
All Photos(6)

Documents

SAB1402125

Sigma-Aldrich

Monoclonal Anti-ATP6V1A, (C-terminal) antibody produced in mouse

clone 4F5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ATP6A1, ATP6V1A1, HO68, VA68, VPP2, Vma1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP6V1A(523)

General description

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain A subunit isoforms and is found in all tissues. Transcript variants derived from alternative polyadenylation exist. (provided by RefSeq)

Immunogen

ATP6V1A (NP_001681, 508 a.a. ~ 617 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TLEVAKLIKDDFLQQNGYTPYDRFCPFYKTVGMLSNMIAFYDMARRAVETTAQSDNKITWSIIREHMGDILYKLSSMKFKDPLKDGEAKIKSDYAQLLEDMQNAFRSLED

Biochem/physiol Actions

The vacuolar-ATPases significantly control the internal pH by maintaining it neutral. ATP6V1A (ATPase H+ transporting V1 subunit A) is known to be associated with the acidification and intracellular trafficking of secretory granules, endosomes, and lysosomes. Mutations in the gene affect the structure and organisation of V-ATPase, glycosylation of proteins, Golgi trafficking. Mutations can also affect the lysosomal function and results in abnormal extracellular matrix homeostasis. ATP6V1A might serve as a prognostic factor in gastric cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and role of V1A subunit of V-ATPases in gastric cancer cells.
Liu P
International Journal of Clinical Oncology, 20(4), 725-735 (2015)
Mutations in ATP6V1E1 or ATP6V1A Cause Autosomal-Recessive Cutis Laxa.
Van Damme T
American Journal of Human Genetics, 100(2), 216-227 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service