Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300910

Sigma-Aldrich

Anti-CTGF (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-CTGF, Anti-Connective tissue growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
IHC
clone:
polyclonal
species reactivity:
human
citations:
1
technique(s):
immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTGF(1490)

General description

Connective tissue growth factor (CTGF), a member of the CCN family of secreted matricellular proteins, regulates fibrosis, angiogenesis, cell proliferation, apoptosis, tumor growth, and metastasis.

Immunogen

CTGF (NP_001892, 139-170)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the Center region of human CTGF.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service