Skip to Content
Merck
All Photos(2)

Key Documents

S5134

Sigma-Aldrich

Scott′s Tap Water Substitute Concentrate

10 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

powder

IVD

for in vitro diagnostic use

concentration

10 ×

technique(s)

titration: suitable

pH

7.0-7.8

application(s)

hematology
histology

storage temp.

room temp

Application

Used as a "blueing reagent" in hematoxylin and eosin staining procedures. Scott′s Tap Water Substitute Concentrate has been used in transferase−mediated dUTP nick end labeling (TUNEL) assay. It has been used in immunohistochemical analysis.

Other Notes

MgSO4 • 7H2O, 200 g/L + NaHCO3, 20 g/L

Preparation Note

Scott′s Tap Water Substitute is prepared by mixing 1 part of Scott′s Tap Water Substitute Concentrate with 9 parts deionized water.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intermittent metronomic drug schedule is essential for activating antitumor innate immunity and tumor xenograft regression
Chen CS, et al.
Neoplasia, 16(1), W22-W27 (2014)
Histophatologic Techniques (2006)
Impact of tumor vascularity on responsiveness to antiangiogenesis in a prostate cancer stem cell-derived tumor model
Zhang, et al.
Molecular Cancer Therapeutics (2013)
Beth G Ashinsky et al.
Acta biomaterialia, 111, 232-241 (2020-05-25)
Tissue-engineered replacement discs are an area of intense investigation for the treatment of end-stage intervertebral disc (IVD) degeneration. These living implants can integrate into the IVD space and recapitulate native motion segment function. We recently developed a multiphasic tissue-engineered disc-like
Kautilya Kumar Jena et al.
The EMBO journal, 37(18) (2018-08-26)
Sequestration of protein aggregates in inclusion bodies and their subsequent degradation prevents proteostasis imbalance, cytotoxicity, and proteinopathies. The underlying molecular mechanisms controlling the turnover of protein aggregates are mostly uncharacterized. Herein, we show that a TRIM family protein, TRIM16, governs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service