Skip to Content
Merck
All Photos(2)

Key Documents

HT110132

Sigma-Aldrich

Eosin Y Solution, Alcoholic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

0.5 % (w/v) in acidified ethanol

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

General purpose cytoplasmic counterstain. Used with hematoxylin and eosin staining.

Other Notes

Certified Eosin Y, 0.5% (w/v) in acidified 90% ethanol

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitlin L Grzeskowiak et al.
Nature communications, 9(1), 2732-2732 (2018-07-18)
Genetic aberrations driving pro-oncogenic and pro-metastatic activity remain an elusive target in the quest of precision oncology. To identify such drivers, we use an animal model of KRAS-mutant lung adenocarcinoma to perform an in vivo functional screen of 217 genetic
Ek Kia Tan et al.
Investigative ophthalmology & visual science, 52(7), 4534-4545 (2011-04-30)
Intrastromal invasion by limbal basal epithelial progenitor cells in explant cultures is associated with epithelial-mesenchymal transition. It remains unclear whether intrastromal invasion is contingent on culturing conditions and whether invaded cells retain their progenitor status and original lineage. Human limbal
Woo-Young Jeon et al.
PloS one, 11(10), e0164833-e0164833 (2016-10-16)
Gumiganghwal-tang is a traditional herbal prescription that is used widely for the treatment of the common cold and inflammatory diseases in Korea and other Asian countries. In this study, we investigated the protective effects of a Gumiganghwal-tang aqueous extract (GGTA)
Helen F Irving-Rodgers et al.
Cell and tissue research, 339(3), 613-624 (2009-12-25)
Despite the mouse being an important laboratory species, little is known about changes in its extracellular matrix (ECM) during follicle and corpora lutea formation and regression. Follicle development was induced in mice (29 days of age/experimental day 0) by injections
Alexia Eliades et al.
The Journal of biological chemistry, 286(31), 27630-27638 (2011-06-15)
Lysyl oxidase (LOX), a matrix cross-linking protein, is known to be selectively expressed and to enhance a fibrotic phenotype. A recent study of ours showed that LOX oxidizes the PDGF receptor-β (PDGFR-β), leading to amplified downstream signaling. Here, we examined

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service