Skip to Content
Merck
All Photos(1)

Documents

S4887

Sigma-Aldrich

D-(+)-Sorbose

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724559
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥99%

form

powder

optical activity

[α]20/D 36 to 44 °, c = 1% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5+,6+/m1/s1

InChI key

BJHIKXHVCXFQLS-PYWDMBMJSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jessie Fernandez et al.
PLoS genetics, 8(5), e1002673-e1002673 (2012-05-10)
Understanding the genetic pathways that regulate how pathogenic fungi respond to their environment is paramount to developing effective mitigation strategies against disease. Carbon catabolite repression (CCR) is a global regulatory mechanism found in a wide range of microbial organisms that
D B Moody et al.
The Journal of experimental medicine, 192(7), 965-976 (2000-10-04)
T cells recognize microbial glycolipids presented by CD1 proteins, but there is no information regarding the generation of natural glycolipid antigens within infected tissues. Therefore, we determined the molecular basis of CD1b-restricted T cell recognition of mycobacterial glycosylated mycolates, including
Zhangliang Zhu et al.
Microbial cell factories, 18(1), 59-59 (2019-03-27)
A novel D-allulose 3-epimerase from Staphylococcus aureus (SaDAE) has been screened as a D-allulose 3-epimerase family enzyme based on its high specificity for D-allulose. It usually converts both D-fructose and D-tagatose to respectively D-allulose and D-sorbose. We targeted potential biocatalysts
Gustavo Bravo Ruiz et al.
Current genetics, 65(5), 1217-1228 (2019-04-26)
Candida auris is a newly emerged pathogenic microbe, having been identified as a medically relevant fungus as recently as 2009. It is one of the most drug-resistant yeast species known to date and its emergence and population structure are unusual.
Marieke Teppner et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(4), 560-569 (2016-01-29)
The nonsteroidal androgen-receptor antagonist flutamide is associated with hepatic injury. Oxidative stress and reactive metabolite formation are considered contributing factors to liver toxicity. Here we have used flutamide as a model drug to study the generation of reactive drug metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service