Skip to Content
Merck
All Photos(1)

Key Documents

85541

Sigma-Aldrich

L-(−)-Sorbose

for biotechnological purposes, ≥98.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724554
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (sum of enantiomers, HPLC)

form

solid

optical activity

[α]20/D −43±2°, c = 5% in H2O

color

colorless

mp

163-165 °C

solubility

H2O: 0.1 g/mL, clear, colorless to very faintly yellow

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O)C(=O)CO

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5+,6+/m0/s1

InChI key

BJHIKXHVCXFQLS-OTWZMJIISA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Sorbose is used as a starting reagent for the commercial biosynthesis of ascorbic acid (vitamin C).

Biochem/physiol Actions

The L enatiomer of sorbose, a ketohexose monosaccharide.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengzhu Li et al.
AMB Express, 8(1), 38-38 (2018-03-15)
Pyranose oxidase (POx) is a homotetrameric flavoprotein that catalyzes the oxidation of pyranose-configured sugars at position C-2 to corresponding 2-ketoaldoses. The wide substrate specificity makes POx potential for application in various biotechnological industries. In the present study we reported the
Weizhu Zeng et al.
Bioresource technology, 318, 124069-124069 (2020-09-12)
The 2-keto-L-gulonic acid (2-KLG) is the direct precursor for industrial vitamin C production. The main biosynthetic method for 2-KLG production is the classical two-step fermentation route. However, disadvantages of this method are emerging, including high consumption of energy, difficulties in
Raquel Barajas-Azpeleta et al.
PLoS genetics, 14(10), e1007440-e1007440 (2018-10-13)
Antimicrobial peptides act as a host defense mechanism and regulate the commensal microbiome. To obtain a comprehensive view of genes contributing to long-term memory we performed mRNA sequencing from single Drosophila heads following behavioral training that produces long-lasting memory. Surprisingly
Praveen Kumar Reddy et al.
Yeast (Chichester, England), 38(3), 222-238 (2020-11-13)
Human fungal pathogen Candida albicans cannot utilize L-sorbose as a sole carbon source. However, chromosome 5 monosomic strains can grow on sorbose as repressors present on this chromosome get diminished allowing the expression of sorbose utilization gene (SOU1) located on
Xiaoyu Shan et al.
Frontiers in bioengineering and biotechnology, 8, 194-194 (2020-04-08)
2-Keto-L-gulonic acid (2-KLG) is the direct precursor for the production of L-ascorbic acid (L-Asc) on industrial scale. Currently, the production of L-Asc in the industry is a two-step fermentation process. Owing to many unstable factors in the fermentation process, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service