Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Streptavidin from
Streptomyces avidinii
CAS Number:
9013-20-1
Products
Brand
Sigma-Aldrich (3)
Physical Form
Recombinant Host
Quality Segment
Application
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S4762
Streptavidin from
Streptomyces avidinii
,
essentially salt-free, lyophilized powder, ≥13 units/mg protein
Expand
View Pricing
85878
Streptavidin from
Streptomyces avidinii
,
affinity purified, lyophilized from 10 mM potassium phosphate, ≥13 U/mg protein
Expand
View Pricing
S0677
Streptavidin from
Streptomyces avidinii
,
recombinant, expressed in
E. coli
, lyophilized powder
Expand
View Pricing