Skip to Content
Merck
All Photos(1)

Key Documents

S4701

Sigma-Aldrich

Suplatast tosylate

≥98% (HPLC)

Synonym(s):

(+/-)-[2-[4-(3-ethoxy-2-hydroxypropoxy)phenylcarbamoyl]-ethyl] dimethylsulfonium p-toluenesulfonate, IPD-1151T, ST

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26SNO4 · C7H7SO3
CAS Number:
Molecular Weight:
499.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CCOCC(O)COc2ccc(NC(=O)CC[S+](C)C)cc2

InChI

1S/C16H25NO4S.C7H8O3S/c1-4-20-11-14(18)12-21-15-7-5-13(6-8-15)17-16(19)9-10-22(2)3;1-6-2-4-7(5-3-6)11(8,9)10/h5-8,14,18H,4,9-12H2,1-3H3;2-5H,1H3,(H,8,9,10)

InChI key

RYVJQEZJUFRANT-UHFFFAOYSA-N

Application

Suplatast tosylate may be used in immune-signaling studies.

Biochem/physiol Actions

Suplatast tosylate is a dimethylsulfonium compound downregulates the expression of histamine H1 receptor gene, suppresses the activity of histidine decarboxylase and regulates IL-4 mRNA. It inhibits the release of histamines and regulates the production of IgE.1,2 Suplatast tosylate is effective in the treatment of mild-to-moderate persistent asthma in children.3
Suplatast tosylate is an anti-allergy, cytokine modulator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fatemeh Mousavi et al.
Bioorganic chemistry, 90, 103037-103037 (2019-06-19)
Capecitabine as a prodrug of 5-Fluorouracil plays an important role in the treatment of breast and gastrointestinal cancers. Herein, in view of the importance of this drug in chemotherapy, interaction mechanism between Capecitabine (CAP) and human serum albumin (HSA) as
Sandhir B Prasad et al.
The American journal of cardiology, 124(3), 325-333 (2019-06-04)
The inter-relationships between minimal and maximal left atrial volume index (LAVI), left ventricular filling pressures and survival have not been well studied. This study aimed to compare LAVImin with LAVImax with respect to (1) relative prognostic value, and (2) correlation
Ana Moreno-Alcázar et al.
Trials, 18(1), 160-160 (2017-04-06)
Up to 60% of patients with bipolar disorder (BD) have a history of traumatic events, which is associated with greater episode severity, higher risk of comorbidity and higher relapse rates. Trauma-focused treatment strategies for BD are thus necessary but studies
Roberta W Scherer et al.
JAMA otolaryngology-- head & neck surgery, 145(7), 597-608 (2019-05-24)
Tinnitus retraining therapy (TRT) is an internationally recognized, but controversial, protocol of uncertain efficacy that uses tinnitus-specific educational counseling (TC) and sound therapy (ST) to reduce the patient's tinnitus-evoked negative reaction to, and awareness of, tinnitus. To compare the efficacy
Flemming K Nielsen et al.
BMC musculoskeletal disorders, 18(1), 510-510 (2017-12-06)
To evaluate whether clinical, radiographic or MRI findings are associated with long term risk for total knee arthroplasty (TKA) in persons with knee osteoarthritis. We performed a follow-up analysis of 100 persons with knee osteoarthritis who participated in a clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service