Skip to Content
Merck
All Photos(1)

Documents

S1452

Sigma-Aldrich

MISSION® siRNA Transfection Reagent

Synonym(s):

MISSION® siRNA Transfection Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106502
NACRES:
NA.51

product line

MISSION®

form

aqueous solution

shipped in

wet ice

storage temp.

2-8°C

General description

The MISSION® siRNA Transfection Reagent is for the transfection of siRNA to achieve ≥90% silencing efficiency for transient knockdown of eukaryotic gene expression.
Effective gene silencing and reduced off target effects are achieved using a low siRNA concentration (1nM). The siRNA Transfection Reagent is provided as a sterile solution, and is compatible with serum and antibiotics.

Other Notes

Gene silencing assay: An A549 cell line stably expressing firefly luciferase was transfected in 24-well plates with 8.4 ng of 21-mer Luciferase siRNA using 2 μL of the MISSION® siRNA Transfection Reagent. Experiments were performed in triplicate in presence of serum. Cell lysates were assessed for luciferase expression 48 hours post-transfection.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F

Flash Point(C)

49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuyen T Dang et al.
Cancers, 12(7) (2020-07-11)
It was recently shown that the 5' to 3' exoribonuclease XRN2 is involved in the DNA damage response. Importantly, loss of XRN2 abrogates DNA double stranded break repair via the non-homologous end-joining pathway. However, the mechanistic details of how XRN2
Tuyen T Dang et al.
International journal of molecular sciences, 21(12) (2020-06-19)
Cellular survival is dependent on the efficient replication and transmission of genomic information. DNA damage can be introduced into the genome by several different methods, one being the act of DNA replication. Replication is a potent source of DNA damage
Ponnuraj Nagendraprabhu et al.
PLoS pathogens, 13(12), e1006779-e1006779 (2017-12-16)
Poxviruses have evolved multiple strategies to subvert signaling by Nuclear Factor κB (NF-κB), a crucial regulator of host innate immune responses. Here, we describe an orf virus (ORFV) virion-associated protein, ORFV119, which inhibits NF-κB signaling very early in infection (≤
Ilaria Guerriero et al.
Oncotarget, 8(12), 19172-19191 (2016-11-24)
Hyperactivation of the PI3K/AKT pathway is observed in most human cancer including lung carcinomas. Here we have investigated the role of miRNAs as downstream targets of activated PI3K/AKT signaling in Non Small Cell Lung Cancer (NSCLC). To this aim, miRNA
Eirini Kefaloyianni et al.
Journal of the American Society of Nephrology : JASN, 30(12), 2370-2383 (2019-11-05)
Sustained activation of EGF receptor (EGFR) in proximal tubule cells is a hallmark of progressive kidney fibrosis after AKI and in CKD. However, the molecular mechanisms and particular EGFR ligands involved are unknown. We studied EGFR activation in proximal tubule

Articles

This brief webinar provides an overview of what transfection is and the methods that are used to introduce DNA or RNA into eukaryotic cells.

Related Content

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

Browse our convenient transfection reagent selection guide to match the best reagent for your specific cell line and application needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service