Skip to Content
Merck
All Photos(1)

Documents

EHUFLUC

Sigma-Aldrich

MISSION® esiRNA

targeting FLUC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

shipped in

ambient

storage temp.

−20°C

General description

EHUFLUC targets Firefly Luciferase. It can be used as a negative control in systems lacking Firefly Luciferase, or a positive knockdown control in systems expressing it.
MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Other Notes

esiRNA cDNA target sequence: GAGCAACTGCATAAGGCTATGAAGAGATACGCCCTGGTTCCTGGAACAATTGCTTTTACAGATGCACATATCGAGGTGGACATCACTTACGCTGAGTACTTCGAAATGTCCGTTCGGTTGGCAGAAGCTATGAAACGATATGGGCTGAATACAAATCACAGAATCGTCGTATGCAGTGAAAACTCTCTTCAATTCTTTATGCCGGTGTTGGGCGCGTTATTTATCGGAGTTGCAGTTGCGCCCGCGAACGACATTTATAATGAACGTGAATTGCTCAACAGTATGGGCATTTCGCAGCCTACCGTGGTGTTCGTTTCCAAAAAGGGGTTGCAAAAAATTTTGAACGTGCAAAAAAAGCTCCCAATCATCCAAAAAATTATTATCATGGATTCTAAAACGGATTACCAGGGATTTCAGTCGATGTACACGTTCGTCACATCTCATCTACCTCCCGGTTTTAATGAATACGATTTTGTGCCAGAGTCCTTCGATAGGGACAAGACAATTGCACTGATCATGAACTCCTCTGGATCTACTGGTCTGCCTAAAGGTGTCGCTCTGCCTCATAGAACTGCCTGCGTGAGATT

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Lee et al.
PloS one, 14(5), e0213116-e0213116 (2019-05-22)
The mitochondrial outer membrane protein Mitochondrial Fission Factor (Mff) plays a key role in both physiological and pathological fission. It is well established that at stressed or functionally impaired mitochondria, PINK1 recruits the ubiquitin ligase Parkin which ubiquitinates Mff and
Ann Hammarstedt et al.
Scientific reports, 8(1), 15757-15757 (2018-10-27)
Adipose tissue dysfunction is considered an important contributor to systemic insulin resistance and Type 2 diabetes (T2D). Recently, a novel family of endogenous lipids, palmitic acid hydroxy stearic acids (PAHSAs), was discovered. These have anti-diabetic and anti-inflammatory effects in mice
Noelia Luna-Peláez et al.
Cell death & disease, 10(8), 548-548 (2019-07-20)
Mutations in NIPBL are the major cause of Cornelia de Lange Syndrome (CdLS). NIPBL is the cohesin-loading factor and has recently been associated with the BET (bromodomains and extra-terminal (ET) domain) proteins BRD2 and BRD4. Related to this, a CdLS-like
Annamaria Morotti et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 35(12), 2423-2431 (2020-08-12)
A role for long non-coding RNAs (lncRNAs) in endocrine cancer pathogenesis is emerging. However, knowledge regarding their expression pattern, correlation with known genetic defects, and clinical implications in parathyroid tumors is still unclear. Here, we profiled 90 known lncRNAs in
Theodora Saridaki et al.
Journal of neurochemistry, 146(4), 474-492 (2018-05-11)
Parkinson's disease can be caused by mutations in the α-synuclein gene and is characterized by aggregates of α-synuclein protein. We have previously shown that over-expression of the small GTPase Rab7 can induce clearance of α-synuclein aggregates. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service