RAB0797
Mouse IgA ELISA Kit
Synonym(s):
IgA, Immunoglobulin A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
shipped in
wet ice
storage temp.
−20°C
General description
The antibody pair provided in this kit recognizes Mouse IgA.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
Immunoglobulin A (IgA) is majorly found in the mucosal secretions. IgA exists as two subclasses, IgA1 and IgA2. IgA is a dimer in animals, but a monomer in humans. IgA interacts with the Fc fragment of IgA receptor (FCAR) to elicit immune response against inhaled pathogens. Secretory IgA (S-IgA) produced by B1 lymphocytes triggers phagocytosis and leucocyte respiratory burst. S-IgA exists as a polymer and is crucial for primary defense. The high levels of IgA in human milk and colostrum may provide immune protection to infants. Deficiency of IgA leads to increased susceptibility to infections. High levels of IgA in kidney is present in IgA nephropathy and IgA myeloma. Autoantibodies of IgA is present in celiac disease.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yiran Wang et al.
Molecular immunology, 112, 305-311 (2019-06-24)
The pathophysiology of type II diabetes (T2D) in patients with normal body-mass index (BMI) remains unclear. In this study, we investigated the circulating CD4+CXCR5+ T cells in lean T2D patients. Compared with age-, gender-, and BMI-matched healthy controls, the lean
Liqing Huang et al.
Probiotics and antimicrobial proteins, 11(3), 946-956 (2018-06-28)
The effects of Enterococcus faecium on growth, intestinal barrier function, and immune response in Escherichia coli O78-challenged broiler chickens were investigated. Three hundred eight 1-day-old Ross male chickens were randomly assigned into three treatment groups: negative control (C), E. coli
T Gupalova et al.
PloS one, 13(5), e0196564-e0196564 (2018-05-05)
Streptococcus agalactiae, or group B streptococcus (GBS), is an important pathogen as it is the leading cause of neonatal deaths due to sepsis, meningitis or bacterial pneumonia. Although the development of an effective and safe GBS vaccine is on the
Function of immunoglobulin A in immunity
Kerr, MA
Gut, 47(6), 751-752 (2000)
IgA function-variations on a theme
Woof, Jenny M and Kerr, Michael A
Immunology, 113(2), 175-177 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service