Skip to Content
Merck
All Photos(1)

Key Documents

RA2DWP-EP

Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges

A-2-DWP, with 2 buckets

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold 2 x 2 DWP
to hold 2 x 4 cell culture plates
to hold 2 x 5 MTP
deepwell plate rotor
with 2 buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 022638564 (NA)
Eppendorf® 5804740009 (EU)

parameter

2,250 × g max. RCF

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5804/5804R/5810/5810R

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service