Skip to Content
Merck
All Photos(1)

Key Documents

RAB0639

Sigma-Aldrich

Human CHI3L1 / Chitinase-3-like Protein 1 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CHI3L1(1116)

General description

The CHI3L1 ELISA kit provides for the Quantitative measurement of Chitinase 3-like in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unnur D Teitsdottir et al.
Alzheimer's research & therapy, 12(1), 92-92 (2020-08-06)
Neuroinflammation has gained increasing attention as a potential contributing factor in the onset and progression of Alzheimer's disease (AD). The objective of this study was to examine the association of selected cerebrospinal fluid (CSF) inflammatory and neuronal degeneration markers with
Pol Andrés-Benito et al.
Aging, 10(9), 2367-2382 (2018-09-15)
Amyotrophic lateral sclerosis (ALS) has variable clinical course and fatal outcome. Since inflammation plays a role in the pathogenesis of ALS, chitinase-3-like protein 1 or YKL40 has been assessed as putative biomarker of disease progression. YKL40 mRNA levels are increased
Sebastian Palmqvist et al.
EMBO molecular medicine, 11(12), e11170-e11170 (2019-11-12)
Failures in Alzheimer's disease (AD) drug trials highlight the need to further explore disease mechanisms and alterations of biomarkers during the development of AD. Using cross-sectional data from 377 participants in the BioFINDER study, we examined seven cerebrospinal fluid (CSF)
Hyun Hee Kim et al.
Annals of laboratory medicine, 38(1), 39-45 (2017-10-27)
Given that YKL-40 is a known marker of inflammation, we sought to determine its association with urinary tract infection (UTI) in febrile children. In total, 44 children aged 0 to 24 months with febrile UTI and 35 age- and sex-matched
Jaco Botha et al.
Nutrition & metabolism, 15, 76-76 (2018-11-06)
Bariatric surgery is a widely adopted treatment for obesity and its secondary complications. In the past decade, microvesicles (MVs) and CD36 have increasingly been considered as possible biomarkers for obesity, the metabolic syndrome (MetSy), type 2 diabetes mellitus (T2DM). Thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service