Skip to Content
Merck
All Photos(1)

Key Documents

R9525

Sigma-Aldrich

RX 821002 hydrochloride

solid, ≥98% (HPLC)

Synonym(s):

2-[2-(2-Methoxy-1,4-benzodioxanyl)]imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O3 · HCl
CAS Number:
Molecular Weight:
270.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white

solubility

H2O: >20 mg/mL

SMILES string

Cl[H].COC1(COc2ccccc2O1)C3=NCCN3

InChI

1S/C12H14N2O3.ClH/c1-15-12(11-13-6-7-14-11)8-16-9-4-2-3-5-10(9)17-12;/h2-5H,6-8H2,1H3,(H,13,14);1H

InChI key

IMPOOMVZVWKSAP-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Selective α2-adrenoceptor antagonist

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Gasparini et al.
Neuroscience, 164(2), 370-379 (2009-09-01)
The blockade of serotoninergic receptors with methysergide or the activation of alpha(2)-adrenoceptors with moxonidine into the lateral parabrachial nucleus (LPBN) increases water and 0.3 M NaCl intake in rats treated with furosemide (FURO) combined with captopril (CAP). In the present
S J Rayment et al.
British journal of pharmacology, 160(7), 1727-1740 (2010-07-24)
We have investigated the distribution of alpha-adrenoceptors in sheep internal anal sphincter (IAS), as a model for the human tissue, and evaluated various imidazoline derivatives for potential treatment of faecal incontinence. Saturation and competition binding with (3)H-prazosin and (3)H-RX821002 were
Mercè Masana et al.
The international journal of neuropsychopharmacology, 14(1), 53-68 (2010-08-13)
The superior efficacy of atypical vs. classical antipsychotic drugs to treat negative symptoms and cognitive deficits in schizophrenia appears related to their ability to enhance mesocortical dopamine (DA) function. Given that noradrenergic (NE) transmission contributes to cortical DA output, we
J Galitzky et al.
British journal of pharmacology, 100(4), 862-866 (1990-08-01)
1. The binding of a new alpha 2-adrenoceptor antagonist, [3H]-RX821002 (2-(2-methoxy-1,4-benzodioxan-2-yl)-2-imidazoline), was investigated in human platelet membranes and compared with [3H]-yohimbine binding parameters. 2. Analysis of kinetic data revealed association and dissociation time courses consistent with a simple biomolecular reaction.
Axel Rominger et al.
Synapse (New York, N.Y.), 64(8), 654-657 (2010-03-26)
To test for alpha(2) adrenergic modulation of dopamine D(2/3) receptor availability in striatum of living mice using the high-affinity ligand [(18)F]fallypride and microPET. Groups of anesthetized mice were pretreated with saline, the alpha(2)-agonist clonidine (1 mg/kg), and the alpha(2)-antagonists RX821002

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service