Skip to Content
Merck
All Photos(1)

Key Documents

Y3125

Sigma-Aldrich

Yohimbine hydrochloride

≥98% (HPLC), powder, α2-adrenergic receptor antagonist

Synonym(s):

17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3 · HCl
CAS Number:
Molecular Weight:
390.90
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Yohimbine hydrochloride, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

288-290 °C (dec.) (lit.)

solubility

H2O: 10 mg/mL

SMILES string

Cl.COC(=O)[C@H]1[C@@H](O)CCC2CN3CCc4c([nH]c5ccccc45)C3CC12

InChI

1S/C21H26N2O3.ClH/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24;/h2-5,12,15,17-19,22,24H,6-11H2,1H3;1H/t12-,15-,17-,18-,19+;/m0./s1

InChI key

PIPZGJSEDRMUAW-VJDCAHTMSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Yohimbine hydrochloride has been:
  • used to study its in vitro antiviral, antibacterial and antifungal activities and cytotoxicity[1]
  • used as a tyramine receptor blocker to oppose the effects of increased tyramine in relation with flight initiation and maintenance deficits in experimental flies[2]
  • used alone and as well as with brimonidine to study the neuroprotective effect of brimonidine in the presence of glutamate-induced neurotoxicity, oxidative stress and hypoxia on in vitro cultures of purified rat retinal ganglion cells[3]

Biochem/physiol Actions

Yohimbine is derived from the Coryanthe yohimbe tree cortex. It is an alkaloid. Yohimbine hydrochloride is an α2-adrenergic receptor antagonist.[4] It blocks the central α2-adrenergic receptors in the brain, thus preventing and reducing the effects of xylazine, an α2-adrenergic agonist. The sedative effects and respiratory depression that accompany xylazine administration are both antagonized and reversed by yohimbine. Yohimbine hydrochloride also produces an antidiuretic effect, and increases heart rate and blood pressure.[5] It is useful in treating erectile dysfunction and promotes sexual functioning.[4]

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MARTINDALE: Martindale?the Extra Pharmacopoeia
Lipman Arthur G
The International journal of pharmacy practice, 2(2), 124-124 (1993)
Susan X Jiang et al.
European journal of pharmacology, 629(1-3), 12-19 (2009-12-05)
Intracellular calcium ([Ca(2+)]i) influx through N-methyl-d-aspartic acid (NMDA) receptors in cortical neurons is central to NMDA receptor-mediated excitotoxicity. Drugs that uncompetitively modulate NMDA receptor-mediated [Ca(2+)]i influx are potential leads for development to treat NMDA receptor-mediated neuronal damage since these drugs
Yohimbine in erectile dysfunction: the facts
Morales, A
International Journal of Impotence Research, 12(S1), S70-S70 (2000)
Flight Initiation and Maintenance Deficits in Flies with Genetically Altered Biogenic Amine Levels
Brembs B, et al.
The Journal of Neuroscience, 27(41), 11122-11131 (2007)
L Pan et al.
Human & experimental toxicology, 39(3), 365-373 (2019-11-28)
Dexmedetomidine (DMED), an alpha-2 adrenoreceptor agonist, has been widely used in regional anesthesia procedures. However, the effect of DMED on local anesthetic cardiotoxicity has not been well delineated. This study consisted of two experiments. In experiment A, 42 Sprague-Dawley (SD)

Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service