Skip to Content
Merck
All Photos(1)

Documents

R4268

Sigma-Aldrich

RNA Sample Loading Buffer

for NA electrophoresis, with ethidium bromide (50 μg/mL)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161703
NACRES:
NA.25

grade

for molecular biology

Quality Level

form

liquid

concentration

1.25 ×

technique(s)

electrophoresis: suitable

foreign activity

RNase, none detected

storage temp.

−20°C
−20°C

Looking for similar products? Visit Product Comparison Guide

General description

RNA loading buffer is used as a tracking dye during RNA electrophoresis. The RNA loading dye has a slight negative charge and will migrate the same direction as RNA, allowing the user to monitor the progress of molecules moving through the gel. The rate of migration varies with gel composition. Dilute 1:3 to 1:6 with sample, heat to 65C for ten minutes and chill on ice before loading.

Application

RNA sample loading buffer is especially formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide. Ethidium bromide is not recommended for gel staining prior to Northern blot detection because the presence of ethidium bromide in agarose gels or the loading buffer can cause poor transfer efficiency.
Suitable for use with formaldehyde-agarose gels used in Northern blotting procedures.
RNA Sample Loading Buffer has been used as a sample loading buffer in northern blot.

Components

Deionized formamide 62.5% (v/v), formaldehyde 1.14 M, bromphenol blue 200 μg/mL, xylene cyanole 200 μg/mL, MOPS-EDTA-sodium acetate at 1.25× working concentration.
RNA loading buffer contains 62.5% deionized formamide, 1.14M formaldehyde, 200 μg/ml bromphenol blue, 200 μg/ml xylene cyanole, and 50 μg/ml ehtidium bromide in MOPS-EDTA-sodium acetate at 1.25x working concentration.

Quantity

Recommended usage: Add 1 volume sample to 2-5 volumes of sample loading buffer and mix well. The sample should be heated to 65 °C for 10 minutes, and then chilled on ice immediately before loading on the gel.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Muta. 2 - Repr. 1B - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Identification and characterization of bicistronic speB and prsA gene expression in the group A Streptococcus.
Ma Y, et al.
Journal of Bacteriology, 188, 7626-7634 (2006)
Erika C von Grote et al.
Molecular bioSystems, 7(1), 150-161 (2010-08-24)
Cytokines are important mediators of the wound healing response. However, sampling of cytokines from the interstitial fluid at a healing wound site in experimental animals is a challenge. Microdialysis sampling is an in vivo collection option for this purpose as
The effect of massive small bowel resection and oral epidermal growth factor therapy on SGLT-1 distribution in rabbit distal remnant.
Chung BM, et al.
Pediatric Research, 55, 19-26 (2004)
Amy E Bryant et al.
Journal of medical microbiology, 68(3), 456-466 (2019-01-25)
Extracellular protein toxins contribute to the pathogenesis of Staphylococcus aureus infections. The present study compared the effects of iclaprim and trimethoprim - two folic acid synthesis inhibitors - with nafcillin and vancomycin on production of Panton-Valentine leukocidin (PVL), alpha haemolysin
Impact of antibiotics on expression of virulence-associated exotoxin genes in methicillin-sensitive and methicillin-resistant Staphylococcus aureus.
Stevens DL, et al.
The Journal of Infectious Diseases, 195, 202-211 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service