Skip to Content
Merck
All Photos(2)

Documents

M8032

Sigma-Aldrich

Mixed Bed Resin

BioReagent, for molecular biology

Synonym(s):

Mixed Bed Resin hydrogen and hydroxide form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.52

grade

DNA grade
for molecular biology

product line

BioReagent

form

beads

pH

<5

foreign activity

DNase, RNase, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

A self-indicating Mixed Bed Resin for deionizing formamide, acrylamide, glyoxal, PEG and urea. The beads change color from blue to gold when exchange capacity is reached. Also, the color indicator turns gold with formamide immediately, so deionization must be checked by measuring conductivity.

Application

Mixed Bed Resin is suitable for deionizing formamide, acrylamide, glyoxal, PEG and urea, on a column or in a batch process.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Hsiu Lin et al.
Langmuir : the ACS journal of surfaces and colloids, 29(25), 7793-7801 (2013-05-25)
Zeta potentials of several polar protic (water, ethylene glycol, and formamide) as well as polar aprotic (dimethyl sulfoxide) liquids were measured in contact with three nonpolar surfaces using closed-cell electroosmosis. The test surfaces were chemisorbed monolayers of alkyl siloxanes, fluoroalkyl
A I Alayash
Free radical biology & medicine, 18(2), 295-301 (1995-02-01)
Chemical modifications of human or bovine hemoglobins are designed to produce proteins that can act as oxygen-carrying blood substitutes. Concerns about the redox reactivity of cell-free hemoglobin and its contribution to tissue-damaging oxygen free radicals has not been fully established.
Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 1-1 (1989)
E G Vilela et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 41(12), 1105-1109 (2009-01-17)
The gut barrier monitors and protects the gastrointestinal tract from challenges such as microorganisms, toxins and proteins that could act as antigens. There is evidence that gut barrier dysfunction may act as a primary disease mechanism in intestinal disorders. The
Glycogen synthase activity in adipose tissue. Methods for freeze-clamping and assay.
H K Ortmeyer
Methods in molecular biology (Clifton, N.J.), 155, 89-96 (2001-04-11)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service