Skip to Content
Merck
All Photos(1)

Key Documents

PZ0255

Sigma-Aldrich

CP-101606 mesylate

≥98% (HPLC)

Synonym(s):

(1S,2S)-1-(4-Hydroxy-phenyl)-2-(4-hydroxy-4-phenylpiperidino)-1-propanol mesylate, (1S,2S)-1-(4-Hydroxyphenyl)-2-(4-hydroxy-4-phenylpiperidin-1-yl)-1-propanol mesylate, CP-101,60 mesylate, CP-101606-27, Traxoprodil mesylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO3 · CH4O3S
CAS Number:
Molecular Weight:
423.52
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

OC1=CC=C([C@H](O)[C@H](C)N2CCC(O)(C3=CC=CC=C3)CC2)C=C1.CS(=O)(O)=O

InChI

1S/C20H25NO3.CH4O3S/c1-15(19(23)16-7-9-18(22)10-8-16)21-13-11-20(24,12-14-21)17-5-3-2-4-6-17;1-5(2,3)4/h2-10,15,19,22-24H,11-14H2,1H3;1H3,(H,2,3,4)/t15-,19+;/m0./s1

InChI key

OZBWUANKVIMZIA-WRRDZZDISA-N

Biochem/physiol Actions

CP-101,606 is an NR2B (N-methyl D-aspartate receptor subtype 2B) subunit antagonist, that can increase dyskinesia. It reduces Ca2+/calmodulin-dependent protein kinase II level, at threonine-286 hyperphosphorylation (pCaMKII-Thr286). It has no effect on the α1-adrenergic receptors. CP-101,606 has the ability to inhibits the habits, stimulated by cocaine. It also possesses hallucinogenic properties.
CP-101606 mesylate (Traxoprodil mesylate) is a potent noncompetitive antagonist of N-methyl-D-aspartate (NMDA) receptors, selective for the NR2B subunit. CP-101606 mesylate has been shown to be neuroprotective in animal models of brain injury and stroke.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the selective antagonist of the NR2B subunit of the NMDA receptor, traxoprodil, on the antidepressant-like activity of desipramine, paroxetine, milnacipran, and bupropion in mice
Stasiuk, et al.
Journal of neural transmission (Vienna, Austria : 1996), 124(3), 387-396 (2017)
Induction and blockade of adolescent cocaine-induced habits
DePoy, et al.
Biological Psychiatry, 81(7), 595-605 (2017)
NR2B antagonist CP-101,606 inhibits NR2B phosphorylation at tyrosine-1472 and its interactions with Fyn in levodopa-induced dyskinesia rat model
Kong, et al.
Behavioural Brain Research, 282, 46-53 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service