Skip to Content
Merck
All Photos(1)

Documents

S8071

Sigma-Aldrich

Salvinorin A

≥98% (HPLC), solid

Synonym(s):

(2S,4aR,6aR,7R,9S,10aS,10bR)-9-(acetyloxy)-2-(3-furanyl)dodechydro-6a,10b-dimethyl-4,10-dioxo-2H-naphtho[2,1-c]pyran-7-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

protect from light

color

white

solubility

DMSO: ≥10 mg/mL
ethanol: ~3 mg/mL

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@@]3(C)[C@@H](C[C@H](OC(C)=O)C(=O)[C@]3([H])[C@@]1(C)C[C@H](OC2=O)c4ccoc4)C(=O)OC

InChI

1S/C23H28O8/c1-12(24)30-16-9-15(20(26)28-4)22(2)7-5-14-21(27)31-17(13-6-8-29-11-13)10-23(14,3)19(22)18(16)25/h6,8,11,14-17,19H,5,7,9-10H2,1-4H3/t14-,15-,16-,17-,19-,22-,23-/m0/s1

InChI key

OBSYBRPAKCASQB-AGQYDFLVSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Salvinorin A was administered to rats to study effects on lactic acid-stimulated stretching.1

Biochem/physiol Actions

Salvinorin A is a potent, non-nitrogenous κ opioid selective receptor agonist. Salvinorin A is isolated from Salvia divinorum. Salvinorin A displays high affinity at both native (Ki=4.3 nm) and cloned (Ki=16 nm) κ -opioid receptors. Preliminary studies suggest that Salvanorin A is chemically unique among the psychtropic drugs and does not bind to any known receptor.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Thomas E Prisinzano
Journal of medicinal chemistry, 56(9), 3435-3443 (2013-04-04)
The neoclerodane diterpene salvinorin A is the major active component of the hallucinogenic mint plant Salvia divinorum Epling and Játiva (Lamiaceae). Since the finding that salvinorin A exerts its potent psychotropic actions through the activation of opioid receptors, the site
John Stogner et al.
The International journal on drug policy, 23(6), 512-521 (2012-04-17)
A plant with dissociative and psychoactive properties began to attract the attention of the media and United States policymakers following a well-publicized suicide in 2006 and reports that the plant served as a 'legal high' and substitute for cannabis. As
Justyna Piekielna et al.
Postepy biochemii, 58(4), 485-491 (2012-01-01)
Salvinorin A (SA) is the main active ingredient of Salvia divinorum, a naturally occurring hallucinogen plant from Mexico. Traditionally, herbal preparations obtained from Salvia were used by the Mazatec Indians for their divination rites and the treatment of gastrointestinal disorders.
Diansan Su et al.
Anesthesia and analgesia, 114(1), 200-204 (2011-11-15)
Cerebral hypoxia/ischemia during infant congenital heart surgery is not uncommon and may induce devastating neurologic disabilities persistent over the lifespan. Hypoxia/ischemia-induced cerebrovascular dysfunction is thought to be an important contributor to neurological damage. No pharmacological agents have been found to
Francesca Guida et al.
Molecular pain, 8, 60-60 (2012-08-24)
Salvinorin A (SA), the main active component of Salvia Divinorum, is a non-nitrogenous kappa opioid receptor (KOR) agonist. It has been shown to reduce acute pain and to exert potent antinflammatory effects. This study assesses the effects and the mode

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service