Skip to Content
Merck
All Photos(2)

Documents

P9178

Sigma-Aldrich

Prochlorperazine dimaleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClN3S Ā· 2C4H4O4
CAS Number:
Molecular Weight:
606.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

solubility

H2O: slightly soluble 0.3 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 2.3 mg/mL
methanol: soluble

originator

GlaxoSmithKline

SMILES string

OC(=O)\C=C/C(O)=O.OC(=O)\C=C/C(O)=O.CN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1

InChI

1S/C20H24ClN3S.2C4H4O4/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)25-20-8-7-16(21)15-18(20)24;2*5-3(6)1-2-4(7)8/h2-3,5-8,15H,4,9-14H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-

InChI key

DSKIOWHQLUWFLG-SPIKMXEPSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Prochlorperazine is an anti-nausea and neuroleptic agent. It is known to cause neuropsychiatric side effects. Prochlorperazine inhibits the action of dopamine in the central nervous system. It acts as an antiemetic drug and prevents vomiting and stimulates sleep. Prochlorperazine is useful as a tranquilizer in the treatment of schizophrenia and nonpsychotic anxiety.
Phenothiazine antipsychotic; D2 dopamine receptor antagonist; antispasmodic

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact. - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Saunders Handbook of Veterinary Drugs - E-Book, 652-652 (2010)
Psychiatric Side Effects of Prescription and Over-the-counter Medications: Recognition and Management, 1946-1946 (1998)
S C Lummis et al.
Neuropharmacology, 36(4-5), 665-670 (1997-04-01)
The effects of a range of phenothiazines were examined on 5-hydroxytryptamine3 (5-HT3) receptors in membranes from NIE-115 neuroblastoma cells using radioligand binding. Chlorpromazine, fluphenazine, perphenazine, trifluoperazine and prochlorperazine inhibited specific binding of both the 5-HT3 receptor antagonist [3H]GR65630 and agonist
Vitaliy E Kalashnikov et al.
Life (Basel, Switzerland), 11(11) (2021-11-28)
A gradual increase in rat soleus muscle electromyographic (EMG) activity is known to occur after 3-4 days of hindlimb suspension/unloading (HS). The physiological significance and mechanisms of such activity of motoneurons under unloading conditions are currently unclear. Since hyperactivity of
Kevin Denis et al.
Nature microbiology, 4(6), 972-984 (2019-03-27)
Bacterial virulence factors are attractive targets for the development of therapeutics. Type IV pili, which are associated with a remarkable array of properties including motility, the interaction between bacteria and attachment to biotic and abiotic surfaces, represent particularly appealing virulence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service