Skip to Content
Merck
All Photos(1)

Documents

P9170

Sigma-Aldrich

Anti-Human IgG (γ-chain specific)−R-Phycoerythrin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (γ-chain specific)−R-PE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:16

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Affinity isolated antigen specific antibody is obtained from goat anti-human IgG antiserum by immunospecific purification which removes essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to the γ-chain of human IgG. Goat anti-human IgG is conjugated to Sigma R-Phycoerythrin (Product No. P8912) by a modification of the method of Kronick Unbound phycoerythrin is removed by gel filtration chromatography
Specificity for the γ-chain of human IgG is determined by Ouchterlony Double Diffusion (ODD). The antibody preparation is specific for human IgG when tested against purified human IgA, IgG, IgM, Bence Jones κ and Bence Jones Lambda myeloma proteins, prior to conjugation
Identity and purity of the antibody is established by immunoelectrophoresis (IEP), prior to conjugation Electrophoresis of the antibody preparation followed by diffusion versus anti-goat IgG and anti-goat whole serum results in single arcs of precipitation.
Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders
Anti-Human IgG (γ-chain specific)-R-Phycoerythrin antibody is specific for human IgG when tested against purified human IgA, IgG, IgM, and Bence Jones κ and λ myeloma proteins.
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. It consists of a gamma (γ) heavy chain in the constant (C) region. The primary structure of the antibody contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and also resides inside the chains.

Immunogen

Developed in goat using purified human IgG as the immunogen.
Purified human IgG

Application

A minimum 1:16 dilution is required for direct immunofluorescent labelling of human peripheral blood lymphocytes. The antibody may be used in fluorescence-activated cell sorting applications.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.1 mM EDTA, 1 mM iodoacetamide, 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Christopher P Stefan et al.
Microorganisms, 9(3) (2021-04-04)
Ebola virus is a continuing threat to human populations, causing a virulent hemorrhagic fever disease characterized by dysregulation of both the innate and adaptive host immune responses. Severe cases are distinguished by an early, elevated pro-inflammatory response followed by a
Gerald Klanert et al.
Scientific reports, 9(1), 8689-8689 (2019-06-20)
High-throughput siRNA screens were only recently applied to cell factories to identify novel engineering targets which are able to boost cells towards desired phenotypes. While siRNA libraries exist for model organisms such as mice, no CHO-specific library is publicly available
Marta de Almeida Santiago et al.
PloS one, 11(12), e0167669-e0167669 (2016-12-10)
The use of antibodies in immunodiagnostic kits generally implies the conjugation of these proteins with other molecules such as chromophores or fluorochromes. The development of more sensitive quality control procedures than spectrophotometry is essential to assure the use of better
Michaela Strotbek et al.
Metabolic engineering, 20, 157-166 (2013-10-23)
MicroRNAs (miRNAs) are short non-coding RNAs that post-transcriptionally regulate the expression of different target genes and, thus, enable engineered gene networks to achieve complex phenotypic changes in mammalian cells. We hypothesized that exploiting this feature of miRNAs could improve therapeutic
Xiaojie Chu et al.
Frontiers in immunology, 14, 1070492-1070492 (2023-02-11)
Ectonucleotide pyrophosphatase/phosphodiesterase 1 (ENPP1) is a type II transmembrane glycoprotein expressed in many tissues. High expression levels of ENPP1 have been observed in many cancer types such as lung cancer, ovarian cancer, and breast cancer. Such overexpression has been associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service