Skip to Content
Merck
All Photos(5)

Key Documents

P1274

Sigma-Aldrich

Poly-L-lysine hydrobromide

suitable for cell culture, Mol wt 70,000-150,000 by viscosity

Synonym(s):

L-Lysine homopolymer hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Poly-L-lysine hydrobromide, mol wt 70,000-150,000 by viscosity

form

solid

Quality Level

mol wt

70,000-150,000 by viscosity

technique(s)

cell culture | mammalian: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.NCCCCC(N)C(O)=O

InChI

1S/C18H38N6O4/c19-10-4-1-7-13(22)16(25)23-14(8-2-5-11-20)17(26)24-15(18(27)28)9-3-6-12-21/h13-15H,1-12,19-22H2,(H,23,25)(H,24,26)(H,27,28)/t13-,14-,15-/m0/s1

InChI key

WBSCNDJQPKSPII-KKUMJFAQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly-L-lysine polymers can be used in promoting cell adhesion to solid substrates, conjugation to methotrexate for increased drug transport, microencapsulation of islets, cell microencapsulation technology, microarray glass slide coating, and chromosomal preparations. Lower molecular weight poly-L-lysine (30,000-70,000) is less viscuous in solution, but higher molecular weight versions provide more attachment sites per molecule.

Poly-L-lysine hydrobromide is used for the following applications:
  • Used for some experiments with REF-52 cells, for coating
  • For electrophysiological experiments, cells were seeded on poly-L-lysine hydrobromide coated or uncoated cover-slips
  • Cell culture of rat neurons
  • Indirect binding of monoclonal antibody via poly-llysine

Biochem/physiol Actions

Poly-L-lysine is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. When it is absorbed to the cell culture surface, poly-L-lysine functions to increase the number of positively charged sites available for cell binding. With cells that can digest poly-L-lysine, poly-D-lysine should be used as the attachment factor.

Components

Poly-L-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-L-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the beta structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Caution

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Preparation Note

This product has a molecular weight of 70,000-150,000. To remove the HBr, dissolve this product in a neutral buffer and dialyze to remove the salts. None of the poly-L-lysine products have been exposed to trifluoroacetic acid and are dialyzed to remove any monomers, dimmers, or trimers, confirmed by thin layer chromatography. In general, to use this product as an attachment factor, add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine, and aseptically coat the surface with 1 mL per 25 cm2 of solution. After 5 minutes, remove the solution through aspiration and thoroughly rinse the surface. Let dry for two hours before introducing cells and medium.

Analysis Note

Molecular weight based on viscosity and is also assayed by MALLS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Kaverina et al.
The Journal of cell biology, 142(1), 181-190 (1998-07-14)
By co-injecting fluorescent tubulin and vinculin into fish fibroblasts we have revealed a "cross talk" between microtubules and early sites of substrate contact. This mutuality was first indicated by the targeting of vinculin-rich foci by microtubules during their growth towards
C F Crouch et al.
Journal of clinical microbiology, 19(3), 388-393 (1984-03-01)
Monoclonal antibodies reactive with three different viral polypeptides were evaluated singly and in combination as the capture antibody(s) in an enzyme-linked immunosorbent assay system for the detection of bovine enteric coronavirus. Similar levels of sensitivity were found for all combinations
Y Kubo
The Journal of physiology, 442, 691-710 (1991-10-01)
1. ATP was puff applied to cells of a mesodermal stem cell line, C3H10T1/2, and the responses were studied by whole-cell patch clamp recording. 2. In 91% of the cells (90/99), K+ current lasting for tens of seconds was observed
Jun Wang et al.
PloS one, 12(7), e0181767-e0181767 (2017-07-22)
Triploid plants are usually highly aborted owing to unbalanced meiotic chromosome segregation, but limited viable gametes can participate in the transition to different ploidy levels. In this study, numerous meiotic abnormalities were found with high frequency in an intersectional allotriploid
Y Kubo
The Journal of physiology, 442, 743-759 (1991-10-01)
1. Electrophysiological and immunohistochemical properties during the early stages of muscle differentiation were studied in two myoblastic cell lines, mouse C2C12 and rat L6, and compared to those in myogenic clonal cells derived from the mouse mesodermal stem cell line

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service