Skip to Content
Merck
All Photos(1)

Key Documents

P8824

Sigma-Aldrich

Polyanetholesulfonic acid sodium salt

BioUltra

Synonym(s):

Sodium polyanetholesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.77

product line

BioUltra

form

powder

impurities

≤0.001% Phosphorus (P)
≤0.1% Insoluble matter

anion traces

chloride (Cl-): ≤0.1%

cation traces

Al: ≤0.0005%
Ca: ≤0.05%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.5%
Mg: ≤0.01%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

C/C=C/C1=CC=C(C=C1)OC.[O-]S(=O)[O-].[Na+]

InChI

1S/C10H12O.Na.H2O3S/c1-3-4-9-5-7-10(11-2)8-6-9;;1-4(2)3/h3-8H,1-2H3;;(H2,1,2,3)/q;+1;/p-2/b4-3+;;

InChI key

JKJBFNAERWARKW-CZEFNJPISA-L

Looking for similar products? Visit Product Comparison Guide

General description

Canonical SMILES: CC=CC1=CC=C(C=C1)OC.[O-]S(=O)[O-].[Na+]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Wasilauskas et al.
Journal of clinical microbiology, 35(3), 588-590 (1997-03-01)
We previously reported an inhibitory effect on Mycobacterium avium-M. intracellulare (MAI) when blood collected and processed with the Isolator system was placed in BACTEC 12B bottles for radiometric monitoring. We sought to identify the specific component(s) of the Isolator lysis-anticoagulant
J Cerda et al.
British journal of experimental pathology, 67(5), 623-628 (1986-10-01)
Levels of plasma fibronectin (Fn) were 63% lower than normal 15 min after the intravenous injection of liquoid (P less than 0.01); 3 h later they were still low but rebounded to 35% above normal (P less than 0.01) by
R Shawar et al.
Journal of clinical microbiology, 28(1), 108-111 (1990-01-01)
Haemophilus ducreyi has traditionally been difficult to identify. We have utilized simple test methods to identify 19 fresh isolates obtained during a recent outbreak of chancroid in Houston and six strains of H. ducreyi from other outbreaks. Tests were performed
Kerry Varettas et al.
Journal of clinical microbiology, 43(9), 4613-4615 (2005-09-08)
The tube coagulase test (TCT) performed directly from positive blood culture bottles has been used to reduce the turnaround time for identifying Staphylococcus aureus. Most reports have shown the test to be specific but often lacking sufficient sensitivity to be
P C Braga et al.
Experimental gerontology, 33(5), 477-484 (1998-10-08)
Phagocytic defensive functions consist of a sequence of events, including migration, phagocytosis, secretion, and the release of reactive oxygen species (ROS). The last of these (also called "oxidative burst") has not received due attention in the elderly, even though it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service