Skip to Content
Merck
All Photos(1)

Key Documents

P8615

Sigma-Aldrich

Phosphoinositide 3-kinase p110γ human

≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution

Synonym(s):

PI3K p110γ human, Phosphatidylinositol 3-kinase p110γ human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.32

recombinant

expressed in baculovirus infected insect cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

~4.5 units/mg protein

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIK3CG(5294)

General description

Phosphatidylinositol 3-kinase p110γ /phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit γ (PIK3CG) gene, with 12 exons spanning 43 kb on genomic DNA, is mapped to human chromosome 7q22.3. PIK3CG belongs to the family of genes coding for enzymes that phosphorylate the 3′-hydroxyl of phosphatidylinositol.

Biochem/physiol Actions

PI 3-kinases have been implicated in diverse cellular responses triggered by mammalian cell surface receptors. Inhibits NF-κB, AP-1, and STAT1α mediated gene expression induced by IFN-γ resulting in inhibition of COX-2 and NOS ll (iNOS) expression.
PI 3-kinases have been implicated in diverse cellular responses triggered by mammalian cell surface receptors. Inhibits NF-κB, AP-1, and STAT1α mediated gene expression induced by IFN-γ resulting in inhibition of COX-2 and NOS ll (iNOS) expression.
Phosphatidylinositol 3-kinase p110γ is implicated in the development of eosinophilic inflammation. Thus, inhibition of PI3Kγ expression can be considered as a potential therapeutic method for treating eosinophil-related diseases, including asthma. Knockdown of PIK3CG gene inhibits the PI3K-Akt/PKB signaling pathway, which is essential for tumorigenesis and the progression of colorectal cancers. Mutation in the gene is associated with the development of autistic disorder.

Unit Definition

1 unit will transfer 1 nmol of phosphate/min, using phosphatidylinositol as substrate.

Physical form

Solution in 10 mM HEPES, pH 7.5, 100 mM NaCl, 0.5 mM MgCl2, 50% glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of INPP1, PIK3CG, and TSC2 gene variants with autistic disorder: implications for phosphatidylinositol signalling in autism
Serajee FJ, et al.
Journal of medical Genetics, 40 (2003)
Genome-wide meta-analyses identifies 7 loci associated with platelet aggregation in response to agonists
Johnson AD, et al.
Nature Genetics, 42, 608-613 (2010)
The effect of pharmacological PI3Kγ inhibitor on eotaxin-induced human eosinophil functions.
Saito Y, et al.
Pulmonary Pharmacology & Therapeutics, 27, 164-169 (2014)
Down-Regulation of PIK3CG, a Catalytic Subunit of Phosphatidylinositol 3-OH Kinase, by CpG Hypermethylation in Human Colorectal Carcinoma
Semba S, et al.
Clinical Cancer Research, 8, 3824-3831 (2002)
B Stoyanov et al.
Science (New York, N.Y.), 269(5224), 690-693 (1995-08-04)
Phosphoinositide-3 kinase activity is implicated in diverse cellular responses triggered by mammalian cell surface receptors and in the regulation of protein sorting in yeast. Receptors with intrinsic and associated tyrosine kinase activity recruit heterodimeric phosphoinositide-3 kinases that consist of p110

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service