Skip to Content
Merck
All Photos(2)

Key Documents

P7962

Sigma-Aldrich

Monoclonal Anti-PSTAIR antibody produced in mouse

clone PSTAIR, ascites fluid

Synonym(s):

Anti-CDC2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PSTAIR, monoclonal

mol wt

antigen 33-34 kDa

contains

15 mM sodium azide

species reactivity

mouse, Japanese quail, human, amago salmon, eel, starfish, lily, carp, onion, goldfish, monkey, Xenopus, ciliates

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:4,000 using a COS-7 cell extract

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDC2(983)
mouse ... Cdc2a(12534)

Related Categories

General description

In every eukaryote examined, CDKs contain an evolutionary conserved 16 amino acid sequence called PSTAIR (EGVPSTAIREISLLKE) which distinguishes them from other protein kinases. The PSTAIR motif is involved in complex formation with cyclins. The availability of monoclonal antibody reacting specifically with the PSTAIR sequence enables the subcellular detection and localization of the various CDKs and examination of substrate interactions, in a variety of organisms.
Monoclonal Anti-PSTAIR (mouse IgG1 isotype) is derived from the PSTAIR hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Cyclin-dependent kinase 1 (CDC2/CDK1) belongs to the Ser/Thr protein kinase family. It is located on human chromosome 10q21.

Immunogen

synthetic 16 amino acid oligopeptide containing the PSTAIR sequence conjugated to BSA.

Application

Monoclonal Anti-PSTAIR antibody produced in mouse has been used in:
  • western blot analysis
  • enzyme-linked immunosorbent assay (ELISA)
  • immunoprecipitation

Mouse monoclonal clone PSTAIR anti-PSTAIR antibody may be used for the localization of cyclin-dependent kinases containing the PSTAIR motif using various immunochemical assays including ELISA, immunoblot and immunoprecipitation.

Biochem/physiol Actions

Cyclin-dependent kinase 1 (CDC2/CDK1) controls various mitotic processes including the reorganization of the cytoskeleton, chromosome segregation, and formation and separation of daughter cells. It is essential for the multiplication of mammalian cell. It participates in tumorigenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Takayama et al.
Scientific reports, 6, 38111-38111 (2016-12-03)
Histone gene expression is regulated in a cell cycle-dependent manner, with a peak at S phase, which is crucial for cell division and genome integrity. However, the detailed mechanisms by which expression of histone genes are tightly regulated remain largely
Martin Převorovský et al.
PloS one, 10(9), e0137820-e0137820 (2015-09-15)
Cbf11 and Cbf12, the fission yeast CSL transcription factors, have been implicated in the regulation of cell-cycle progression, but no specific roles have been described and their target genes have been only partially mapped. Using a combination of transcriptome profiling
Héloïse Chassé et al.
PloS one, 11(3), e0150318-e0150318 (2016-03-11)
The cyclin B/CDK1 complex is a key regulator of mitotic entry. Using PP242, a specific ATP-competitive inhibitor of mTOR kinase, we provide evidence that the mTOR signalling pathway controls cyclin B mRNA translation following fertilization in Sphaerechinus granularis and Paracentrotus
Martin Převorovský et al.
Cell cycle (Georgetown, Tex.), 15(22), 3082-3093 (2016-10-22)
For every eukaryotic cell to grow and divide, intricately coordinated action of numerous proteins is required to ensure proper cell-cycle progression. The fission yeast Schizosaccharomyces pombe has been instrumental in elucidating the fundamental principles of cell-cycle control. Mutations in S.
Margit Menges et al.
The Plant cell, 18(4), 893-906 (2006-03-07)
The G1-to-S-phase transition is a key regulatory point in the cell cycle, but the rate-limiting component in plants is unknown. Overexpression of CYCLIN D3;1 (CYCD3;1) in transgenic plants increases mitotic cycles and reduces endocycles, but its effects on cell cycle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service