Skip to Content
Merck
All Photos(1)

Key Documents

P5172

Sigma-Aldrich

Prostaglandin D2

≥95%, synthetic

Synonym(s):

(5Z,9α,13E,15S)-9,15-Dihydroxy-11-oxoprosta-5,13-dien-1-oic-acid, PGD2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
Beilstein:
2170623
MDL number:
UNSPSC Code:
12352211
eCl@ss:
42020658
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[H][C@]1(C\C=C/CCCC(O)=O)[C@@H](O)CC(=O)[C@]1([H])\C=C\[C@@H](O)CCCCC

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-18,21-22H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,18-/m0/s1

InChI key

BHMBVRSPMRCCGG-OUTUXVNYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Primary prostaglandin in brain; induces inflammation; stimulates adenylyl cyclase.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahisa Murata et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(13), 5205-5210 (2013-03-13)
We investigated the role of prostaglandin D2 (PGD2) signaling in acute lung injury (ALI), focusing on its producer-effector interaction in vivo. Administration of endotoxin increased edema and neutrophil infiltration in the WT mouse lung. Gene disruption of hematopoietic PGD synthase
E E Nishizawa et al.
Prostaglandins, 9(1), 109-121 (1975-01-01)
Prostaglandin D2 was found to be a potent inhibitor of platelet aggregation. Aggregation of human platelets by ADP, collagen and prostaglandin G2 was inhibited more strongly by PGD2 than by PGE1. Although ADP-induced aggregation of rabbit platelets was inhibited more
Hana Sarashina et al.
Journal of immunology (Baltimore, Md. : 1950), 192(1), 459-465 (2013-12-04)
The effects of PGD2 are extremely context dependent. It can have pro- or anti-inflammatory effects in clinically important pathological conditions. A greater mechanistic insight into the determinants of PGD2 activity during inflammation is thus required. In this study, we investigated
Alon Mantel et al.
The Journal of investigative dermatology, 132(4), 1103-1110 (2011-12-16)
Aldo-keto reductase 1C3 (AKR1C3) has been shown to mediate the metabolism of sex hormones and prostaglandin D(2) (PGD(2)), a lipid mediator that promotes skin inflammation in atopic dermatitis (AD). As both have a role in skin function and pathology, we
L S Wolfe et al.
Journal of neurochemistry, 53(1), 64-70 (1989-07-01)
In homogenates of rat cerebral neocortex prostaglandin D2 (PGD2) was found to be quantitatively the main PG biosynthesized by a cytosolic PGD synthetase from endogenously released arachidonic acid. Amounts of 628 ng/g wet weight were found after 30-min incubation periods

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service