Skip to Content
Merck
All Photos(1)

Key Documents

P4306

Sigma-Aldrich

Platelet-Derived Growth Factor-BB human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

PDGF-BB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Platelet-Derived Growth Factor-BB human, PDGF-BB, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1.0-3.0 ng/mL EC50

quality

endotoxin tested

mol wt

24.3 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PDGFB(5155)

Looking for similar products? Visit Product Comparison Guide

General description

Platelet-Derived Growth Factor (PDGF), a cationic polypeptide, is an important serum mitogen predominantly found in cells of mesenchymal origin. PDGF is mapped to human chromosome 7p22.3 and is expressed in smooth muscle cells, endothelial cells and macrophages. PDGF exists as hetero and homodimers : PDGF-AB FDGF-AA, PDGF-BB, PDGF-CC and PDGF-DD.

Application

Platelet-Derived Growth Factor-BB human has been used in induction of DNA synthesis and proliferation in low-concentration serum.

Biochem/physiol Actions

Platelet-derived growth factor (PDGF) is a potent chemoattractant and activator of neutrophils, monocytes, and fibroblasts. PDGF elicits multifunctional actions in a variety of cells. It is mitogenic to mesoderm-derived cells, such as dermal and tendon fibroblasts, vascular smooth muscle cells, glial cells and chondrocytes. PDGF-BB isomer is highly expressed in tumor, mediating oncogenic pericyte recruitment and is is a key regulator of angiogenesis. PDGF-BB controls bone homeostasis, repair and regeneration. It increases the synthesis of phospholipids, cholesterol esters, glycogen and prostaglandins and modulates LDL receptor binding.

Other Notes

Dimer of the B chain of human PDGF.

Physical form

Lyophilized from a 0.2 μm filtered solution.

Analysis Note

The biological activity is measured by the dose-dependent stimulation of thymidine uptake in BALB/c 3T3 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The platelet derived growth factor-B polymorphism is associated with risk of severe fever with thrombocytopenia syndrome in Chinese individuals
Zhang XA, et al.
Oncotarget, 7(22), 33340-33349 (2016)
A review of the effects of insulin-like growth factor and platelet derived growth factor on in vivo cartilage healing and repair.
Schmidt MB, et al.
Osteoarthritis and Cartilage / OARS, Osteoarthritis Research Society, 14.5, 403-412 (2006)
Growth factors in the treatment of early osteoarthritis
Civinini R, et al.
Clinical Cases in Mineral and Bone Metabolism : The Official Journal of the Italian Society of Osteoporosis, Mineral Metabolism, and Skeletal Diseases, 10.1, 26-26 (2013)
Overexpression of platelet-derived growth factor-BB increases tumor pericyte content via stromal-derived factor-1a/CXCR4 axis
Song N, et al.
Cancer Research, 69.15, 6057-6064 (2009)
The role of growth factors in wound healing
David L, et al.
The Surgical Clinics of North America, 77.3, 575-586 (1997)

Articles

Platelet-Derived Growth Factor is the principal mitogen found in mammalian serum and is released from platelets during clot formation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service