Skip to Content
Merck
All Photos(1)

Key Documents

P4105

Sigma-Aldrich

Pyruvate Oxidase from Aerococcus sp.

lyophilized powder, ≥35 units/mg protein (biuret)

Synonym(s):

Pyruvate: oxygen oxidoreductase (phosphorylating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Aerococcus spp.)

Quality Level

form

lyophilized powder

specific activity

≥35 units/mg protein (biuret)

storage temp.

−20°C

Application

Pyruvate Oxidase (PoxB) converts pyruvate directly to acetate and CO2. It is used to study pyruvate metabolism. It is used to study aerobic metabolism of bacterium, such as Lactobacillus plantarumand Streptococcus pneumoniae.
Pyruvate Oxidase from Aerococcus sp. has been used in the amplex Red-based fluorescence assay for pyruvate. It is suitable for use in the preparation of biosensor for determination of thiamine (vitamin B1), and as a component in the constant part of the multi-enzyme biocatalytic cascade for the determination of biomarkers for traumatic brain injury (TBI) and soft tissue injury (STI).

Biochem/physiol Actions

Pyruvate Oxidase consists of four subunits with identical molecular weights. PoxB reacts with certain aldehydes and phosphate can be replaced by arsenate. Oxygen as well as several artificial compounds can function as electron acceptors. Pyruvate Oxidase is activated by phospholipids as well as monomeric and micellar amphiphiles.
Pyruvate Oxidase oxidises pyruvate to form acetate, hydrogen peroxide and carbon dioxide. Pyruvate oxidase requires flavin adenine dinucleotide (FAD), thiamine pyrophosphate (TPP) and magnesium as cofactors for its catalytic activity. Thiamine activates pyruvate oxidase activity.

Unit Definition

One unit will produce 1.0 μmole of H2O2 per min during the conversion of pyruvate and phosphate to acetylphosphate and CO2 at pH 6.7 at 37 °C.

Physical form

Lyophilized powder containing buffer salt and sugar

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multi-enzyme logic network architectures for assessing injuries: digital processing of biomarkers
Halamek J, et al.
Molecular Biosystems, 6(12), 2554-2560 (2010)
B Sedewitz et al.
Journal of bacteriology, 160(1), 462-465 (1984-10-01)
Under aerobic growth conditions Lactobacillus plantarum produced acetic acid in addition to lactic acid. It was found that lactic acid was predominantly produced at first, and then when the carbohydrate was nearly exhausted, lactic acid was metabolized further to acetic
Activation of pyruvate oxidase by monomeric and micellar amphiphiles.
R Blake et al.
The Journal of biological chemistry, 253(6), 1963-1971 (1978-03-25)
B Sedewitz et al.
Journal of bacteriology, 160(1), 273-278 (1984-10-01)
Pyruvate oxidase (EC 1.2.3.3) was isolated and characterized from Lactobacillus plantarum. The enzyme catalyzes the oxidative decarboxylation of pyruvate in the presence of phosphate and oxygen, yielding acetyl phosphate, carbon dioxide, and hydrogen peroxide. This pyruvate oxidase is a flavoprotein
A sensitive fluorimetric assay for pyruvate
Zhu A, et al.
Analytical Biochemistry, 396(1), 146-151 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service