Skip to Content
Merck
All Photos(1)

Documents

90101

Sigma-Aldrich

Ampliflu Red

for fluorescence, ≥98.0% (HPLC)

Synonym(s):

10-Acetyl-3,7-dihydroxyphenoxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NO4
CAS Number:
Molecular Weight:
257.24
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

grade

for fluorescence

Assay

≥98.0% (HPLC)

form

powder

fluorescence

λex 571 nm; λem 585 nm in DMSO

storage temp.

2-8°C

SMILES string

CC(=O)N1c2ccc(O)cc2Oc3cc(O)ccc13

InChI

1S/C14H11NO4/c1-8(16)15-11-4-2-9(17)6-13(11)19-14-7-10(18)3-5-12(14)15/h2-7,17-18H,1H3

InChI key

PKYCWFICOKSIHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ampliflu Red has been used to study mitochondrial abnormalities and oxidative stress related to Parkinson’s Disease. Ampliflu Red assay was suitable as an alternative indicator assay for detecting the inhibition of hTPO (human thyroid peroxidase) activity. Phospholipase D, an essential cellular process ingredient, used Ampliflu Red (10-Acetyl-3,7-dihydroxyphenoxazine) to determine its activity.Ampliflu Red is used for the following applications:
  • Quantification of inflammatory mediators
  • Determination of free and esterified cholesterol contents
  • Evaluation of compounds as substrates for GABA-AT compounds

Legal Information

Ampliflu is a trademark of Sigma-Aldrich Co. LLC

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuo Sugiyama et al.
PloS one, 9(4), e94460-e94460 (2014-04-11)
Most of experiments for HCV infection have been done using lytic infection systems, in which HCV-infected cells inevitably die. Here, to elucidate metabolic alteration in HCV-infected cells in a more stable condition, we established an HCV-persistently-infected cell line, designated as
Thais Soprani Ayala et al.
Scientific reports, 9(1), 11447-11447 (2019-08-09)
Macrophages may be a crucial aspect of diabetic complications associated with the inflammatory response. In this study, we examined how hyperglycaemia, a common aspect of diabetes, modulates bone marrow-derived macrophages (BMDMs) under an inflammatory stimulus. To perform this study, BMDMs
Yu Ji et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(63), 16865-16872 (2018-08-29)
Cetyl-trimethylammonium bromide (CTAB) is a widely used cationic surfactant that is biodegradable in nature. CTAB biodegradation requires hydroxylation in the first step, which is rate-limiting and crucial for solubility in water. In this study, the OmniChange multi-site mutagenesis method was
Maria Luisa Di Paolo et al.
Journal of enzyme inhibition and medicinal chemistry, 34(1), 740-752 (2019-03-05)
Fourteen polyamine analogues, asymmetric or symmetric substituted spermine (1-9) or methoctramine (10-14) analogues, were evaluated as potential inhibitors or substrates of two enzymes of the polyamine catabolic pathway, spermine oxidase (SMOX) and acetylpolyamine oxidase (PAOX). Compound 2 turned out to
Marek Krzystanek et al.
Acta biochimica Polonica, 57(3), 369-372 (2010-09-14)
Phospholipase D plays a key role in the biosynthesis of phosphatidic acid, a second messenger involved in essential cellular processes. Oleate-activated phospholipase D was the first mammalian phospholipase D isoform to be discovered but is the least known. The study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service