Skip to Content
Merck
All Photos(2)

Documents

P3017

Sigma-Aldrich

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine

≥95.5% (GC), ≥98% (TLC)

Synonym(s):

(7R,17Z)-4-Hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-3,5,8-trioxa-4-phosphahexacos-17-en-1-aminium 4-oxide, inner salt, 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 2-oleoyl-1-palmitoyl, L-β-Oleoyl-γ-palmitoyl-α-lecithin, PC(16:0/18:1(9Z)), PC(16:0/18:1), PC(16:0/18:1w9), POPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
Beilstein:
4173237
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95.5% (GC)
≥98% (TLC)

form

powder

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20+/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-RYDYYDTQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine (POPC) mimics mammalian phospholipid composition. It comprises a saturated chain (palmitic acid) in the sn-1 position and unsaturated chain (oleic acid) in the sn-2 position,. It is an excellent synthetic substitute for EPC. POPC is associated with alveoli membranes and is a lung surfactant.
Non-pyrogenic, well-defined liposomes, loaded with a molecule of choice, are formed by a single hydration step.

Application

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine has been used:
  • in the liposome preparation for lipid binding assay of monogalactosyldiacylglycerol synthase (MGD1) activity in plant samples
  • in the preparation of liposome for fusion with human umbilical vein endothelial cells (HUVEC) to monitor interleukin1 induced clotting process
  • with trioleoylglycerol for phase separation studies

The effect of non-ionic detergent on liposomes was studied with giant POPC liposomes. Increasing concentrations of detergent cause tubular protrusions, then indentations, to develop on spherical liposomes. At higher detergent concentrations, the liposomes burst.

Biochem/physiol Actions

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine (POPC) reduces air-liquid interface associated surface tension. It is effective in alleviating alveolar collapse and pulmonary edema. POPC is typically considered one of the model lipids for biophysical experiments. It undergoes oxidative decomposition upon ozone exposure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Qiao et al.
The journal of physical chemistry. B, 119(44), 14188-14199 (2015-10-16)
Ambient ozone is a common pollutant in the atmosphere that has an extremely high oxidative ability, can dramatically change the structure and functionality of biomolecules, and is harmful to public health. However, the knowledge about the influence of low-level ozone
Peter van Hoogevest et al.
European journal of lipid science and technology : EJLST, 116(9), 1088-1107 (2014-11-18)
In pharmaceutical formulations, phospholipids obtained from plant or animal sources and synthetic phospholipids are used. Natural phospholipids are purified from, e.g., soybeans or egg yolk using non-toxic solvent extraction and chromatographic procedures with low consumption of energy and minimum possible
Neil R Haria et al.
Biochimica et biophysica acta, 1838(4), 1169-1179 (2014-01-21)
Membrane fusion is critical to eukaryotic cellular function and crucial to the entry of enveloped viruses such as influenza and human immunodeficiency virus. Influenza viral entry in the host cell is mediated by a 20-23 amino acid long sequence, called
Maik Hadorn et al.
Scientific reports, 1, 168-168 (2012-02-23)
Today, free-standing membranes, i.e. liposomes and vesicles, are used in a multitude of applications, e.g. as drug delivery devices and artificial cell models. Because current laboratory techniques do not allow handling of large sample sizes, systematic and quantitative studies on
M Mijajlovic et al.
Colloids and surfaces. B, Biointerfaces, 104, 276-281 (2013-01-22)
Lipid vesicles have received significant attention in areas ranging from pharmaceutical and biomedical engineering to novel materials and nanotechnology. Microfluidic-based synthesis of liposomes offers a number of advantages over the more traditional synthesis methods such as extrusion and sonication. One

Articles

Read our article about how the Nanodisc system allows for structural studies of membrane proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service