Skip to Content
Merck
All Photos(1)

Documents

42773

Sigma-Aldrich

2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine

≥99.0% (TLC)

Synonym(s):

PC, (7R,17Z)-4-Hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-3,5,8-trioxa-4-phosphahexacos-17-en-1-aminium 4-oxide, inner salt, 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 2-oleoyl-1-palmitoyl, L-β-Oleoyl-γ-palmitoyl-α-lecithin, PC(16:0/18:1(9Z)), PC(16:0/18:1), PC(16:0/18:1w9), POPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
Beilstein:
4173237
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Quality Level

Assay

≥99.0% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C\CCCCCCCC

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20+/t40-/m1/s1

InChI key

WTJKGGKOPKCXLL-RYDYYDTQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-pyrogenic, well-defined liposomes, loaded with a molecule of choice, are formed by a single hydration step.

Application

The effect of non-ionic detergent on liposomes was studied with giant POPC liposomes. Increasing concentrations of detergent cause tubular protrusions, then indentations, to develop on spherical liposomes. At higher detergent concentrations, the liposomes burst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil R Haria et al.
Biochimica et biophysica acta, 1838(4), 1169-1179 (2014-01-21)
Membrane fusion is critical to eukaryotic cellular function and crucial to the entry of enveloped viruses such as influenza and human immunodeficiency virus. Influenza viral entry in the host cell is mediated by a 20-23 amino acid long sequence, called
Ivana Malcova et al.
The Journal of biological chemistry, 296, 100607-100607 (2021-04-01)
The respiratory pathogens Bordetella pertussis and Bordetella bronchiseptica employ a type III secretion system (T3SS) to inject a 69-kDa BteA effector protein into host cells. This effector is known to contain two functional domains, including an N-terminal lipid raft targeting
Hsin-Hui Shen et al.
Nature communications, 5, 5078-5078 (2014-10-25)
In biological membranes, various protein secretion devices function as nanomachines, and measuring the internal movements of their component parts is a major technological challenge. The translocation and assembly module (TAM) is a nanomachine required for virulence of bacterial pathogens. We
M Mijajlovic et al.
Colloids and surfaces. B, Biointerfaces, 104, 276-281 (2013-01-22)
Lipid vesicles have received significant attention in areas ranging from pharmaceutical and biomedical engineering to novel materials and nanotechnology. Microfluidic-based synthesis of liposomes offers a number of advantages over the more traditional synthesis methods such as extrusion and sonication. One
Rodrigo M Cordeiro
Biochimica et biophysica acta, 1838(1 Pt B), 438-444 (2013-10-08)
Reactive oxygen species (ROS) are involved in biochemical processes such as redox signaling, aging, carcinogenesis and neurodegeneration. Although biomembranes are targets for reactive oxygen species attack, little is known about the role of their specific interactions. Here, molecular dynamics simulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service