Skip to Content
Merck
All Photos(1)

Key Documents

P0043

Sigma-Aldrich

PNU-120596

≥98% (HPLC)

Synonym(s):

N-(5-Chloro-2,4-dimethoxyphenyl)-N′-(5-methyl-3-isoxazolyl)-urea, NSC 216666

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14ClN3O4
CAS Number:
Molecular Weight:
311.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

O=C(NC1=NOC(C)=C1)NC2=CC(Cl)=C(OC)C=C2OC

InChI

1S/C13H14ClN3O4/c1-7-4-12(17-21-7)16-13(18)15-9-5-8(14)10(19-2)6-11(9)20-3/h4-6H,1-3H3,(H2,15,16,17,18)

InChI key

CEIIEALEIHQDBX-UHFFFAOYSA-N

Biochem/physiol Actions

An allosteric modulator of α7 nicotinic receptors, N-(5-chloro-2,4-dimethoxyphenyl)-N′-(5-methyl-3-isoxazolyl)-urea (PNU-120596), causes conformational changes in the extracellular ligand binding domain similar to those caused by acetylcholinePNU-120596 is a positive allosteric modulator selective for the α7 nicotinic acetylcholine receptor. PNU-120596 produces no detectable change in currents mediated by α4β2, α3β4, α9α10 nAChRs. It increases channel mean open time, but does not affect ion selectivity. It does not bind at the agonist binding site, but induces conformational changes similar to the natural effector.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Morel et al.
Molecular psychiatry, 23(7), 1597-1605 (2017-11-21)
Epidemiological studies report strong association between mood disorders and tobacco addiction. This high comorbidity requires adequate treatment but the underlying mechanisms are unknown. We demonstrate that nicotine exposure, independent of drug withdrawal effects, increases stress sensitivity, a major risk factor
Marta Quadri et al.
Molecular pharmacology, 95(1), 43-61 (2018-10-24)
B-973 is an efficacious type II positive allosteric modulator (PAM) of α7 nicotinic acetylcholine receptors that, like 4BP-TQS and its active isomer GAT107, can produce direct allosteric activation in addition to potentiation of orthosteric agonist activity, which identifies it as
Kateryna Uspenska et al.
The international journal of biochemistry & cell biology, 99, 226-235 (2018-04-29)
Mitochondrial nicotinic acetylcholine receptors (nAChRs) regulate the early stage of mitochondria-driven apoptosis, including cytochrome c release. Mitochondrial nAChR signaling is mainly mediated by intra-mitochondrial kinases, in an ion-independent manner. To determine the relationship between specific nAChR subtypes and mitochondrial kinases
Kateryna Uspenska et al.
Neuroscience letters, 656, 43-50 (2017-07-13)
Several nicotinic acetylcholine receptor (nAChR) subtypes are expressed in mitochondria to regulate the internal pathway of apoptosis in ion channel-independent manner. However, the mechanisms of nAChR activation in mitochondria and targeting to mitochondria are still unknown. Nicotine has been shown
Jean-Rémi Godin et al.
Brain, behavior, and immunity, 87, 286-300 (2019-12-25)
Nicotinic acetylcholine receptors (nAChRs) are best known to function as ligand-gated ion channels in the nervous system. However, recent evidence suggests that nicotine modulates inflammation by desensitizing non-neuronal nAChRs, rather than by inducing channel opening. Silent agonists are molecules that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service