Skip to Content
Merck
All Photos(1)

Key Documents

O9890

Sigma-Aldrich

5Z-7-Oxozeaenol

≥98% (HPLC), powder, TAK1 inhibitor

Synonym(s):

C 292, F 152, FR148083, L 783279, LL-Z1640-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O7
CAS Number:
Molecular Weight:
362.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

5Z-7-Oxozeaenol, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(O)c2C(=O)O[C@@H](C)CC=CC(=O)[C@@H](O)[C@@H](O)C\C=C\c2c1

InChI

1S/C19H22O7/c1-11-5-3-7-14(20)18(23)15(21)8-4-6-12-9-13(25-2)10-16(22)17(12)19(24)26-11/h3-4,6-7,9-11,15,18,21-23H,5,8H2,1-2H3/b6-4+,7-3-/t11-,15-,18+/m0/s1

InChI key

NEQZWEXWOFPKOT-BYRRXHGESA-N

Application

5Z-7-Oxozeaenol has been used:
  • as transforming growth factor β-activated kinase 1 (TAK1) inhibitor to treat human colon cancer cells[1]
  • as TAK1-specific inhibitor to provide intraperitoneal injection to mice for aortic banding[2]
  • as TAK1 inhibitor to treat cells for luciferase reporter assay[3]
  • to test growth and differentiation factor 2 (GDF2)-mediated anoikis[4]

Biochem/physiol Actions

5Z-7-oxozeaenol is a potent ATP-competitive irreversible inhibitor of ERK2 (IC50 = 80 nM), TAK1 (MEKK7), MKK7, and MEK1, which all contain a common cysteine residue in the ATP-binding site. 5Z-7-oxozeaenol has no activity against other MAP kinases. 5Z-7-oxozeaenol is also anti-inflammatory.
The anti-inflammatory effects of 5Z-7-oxozeaenol may affect cardiac hypertrophy.[2]

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the MAPKKKs and MAPKs pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Wang et al.
Journal of cellular and molecular medicine, 25(14), 6584-6601 (2021-06-03)
Gastric cancer (GC) is the most frequent digestive system malignant tumour and the second most common cause of cancer death globally. Cancer stem cell (CSC) is a small percentage of cancer cells in solid tumours that have differentiation, self-renewal and
Cytosolic Internalization of Anti-DNA Antibodies by Human Monocytes Induces Production of Pro-Inflammatory Cytokines Independently of the Tripartite Motif-Containing 21 (TRIM21)-Mediated Pathway
Park H, et al.
Frontiers in Immunology, 9 (2018)
Epigenetic regulation of GDF2 suppresses anoikis in ovarian and breast epithelia
Varadaraj A, et al.
Neoplasia, 17(11), 826-838 (2015)
Li Bo et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 917-924 (2016-10-21)
Resistance to taxol represents a major obstacle for long-term remission in ovarian cancer. Transforming Growth Factor-β-Activated Kinase 1 (TAK1) is a critical component in immune response pathway. However, the role of TAK1 in the development of chemoresistance in ovarian cancer
Disruption of thioredoxin metabolism enhances the toxicity of transforming growth factor beta-activated kinase 1 (TAK1) inhibition in KRAS-mutated colon cancer cells
Hrabe JE, et al.
Redox Biology, 5, 319-327 (2015)

Articles

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

The extracellular signal regulated kinase (ERK1 and ERK2) pathways are activated by mitogens and play an important role in controlling cell growth and differentiation.

See All

Questions

  1. En el CoA de este producto (5Z-7-Oxozeaenol) la concentración en DMSO aparece expresada en MML. ¿Podrían aclararme qué significan esas unidades?¿Masa x vol?¿mmol/L?Porque no consigo encontrar ninguna referencia de concentraciones expresadas en MML.Gracias

    1 answer
    1. This product is analyzed for solubility in DMSO at ≥ 10.0 mg/ mL on a lot to lot basis. The MML references the solubility in mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service