Skip to Content
Merck
All Photos(1)

Key Documents

5.38120

Sigma-Aldrich

NLRP3 Inhibitor, MCC950

Synonym(s):

NLRP3 Inhibitor, MCC950, N-(1,2,3,5,6,7-Hexahydro- s-indacen-4-ylcarbamoyl)-4-(2-hydroxy-2-propanyl)-2-furansulfonamide, Sodium, MCC-950, MCC950, Cytokine Release Inhibitory Drug 3, CRID 3, CP-456,773

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N2O5S · Na
CAS Number:
Molecular Weight:
426.46
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL
DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na+].[S](=O)(=O)(N=C(Nc2c3c(cc4c2CCC4)CCC3)[O-])c1[o]cc(c1)C(O)(C)C

InChI

1S/C20H24N2O5S.Na/c1-20(2,24)14-10-17(27-11-14)28(25,26)22-19(23)21-18-15-7-3-5-12(15)9-13-6-4-8-16(13)18;/h9-11,24H,3-8H2,1-2H3,(H2,21,22,23);/q;+1/p-1

InChI key

LFQQNXFKPNZRFT-UHFFFAOYSA-M

General description

MCC950 is an antagonist ofInterleukin-1 beta (IL-1β). It interacts with both the inactive as well as active forms of NOD-like receptor protein 3 (NLRP3) and targets the Walker B motif within the NLRP3 NACHT domain.[1] A cell-permeable, bioavailable, non-toxic sulfonylurea-derived compound that selectively interacts with the NLRP3 inflammasome and prevents its activation in a reversible manner with no effect on NLRC4 and NLRP1. Dose-dependently reduces IL-1β production (IC50 = 7.5 & 8.1 nM in LPS & ATP-treated BMDMs & HMDMs, respectively) with minimal effect on IL-1α & TNF-α. Specifically, it blocks NLRP3-dependent pyroptotic cell death by inhibiting caspase-1 &-11 activation, IL-1β processing, and ASC oligomerization. Does neither block K+ efflux, Ca2+ flux, or NLRP3-ASC interactions nor inhibits NLRC4, AIM2, TLR signaling, or NLRP3 priming. Effectively suppresses T cell responses and IL-1β& IL-6 secretion, reduces the severity of EAE, and rescues neonatal lethality in a mouse model of CAPS (10 mg/kg, i.p., q.d., & 20 mg/kg, i.p., every other day, respectively). Displays an attractive PK profile with desirable microsomal stability and minimal liability towards CYP450 isozymes (<15% inhibition at 10 µM) & hERG (IC50 >30 µM). It has demonstrated effectiveness against metabolic diseases, autoimmune diseases, cardiovascular diseases, and other conditions.[1]

Application

MCC950 has been used in:

  • hemolysis-affected mice to study the involvement of the NLRP3 inflammasome in kidney function injury.[2]
  • MCC950-mediated inflammasome inhibition assays.[3]
  • testing synergistic effect of pharmacologic inhibition of NLRP3 inflammasome in combination with immune checkpoint inhibitors (ICI) immunotherapy in tumor-bearing animals.[3]
  • studying the role of NLRP3 inflammasome in the development of endothelial dysfunction in early sepsis in mice.[4]

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NLRP3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a sodium salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pleasenote that the molecular weight for this compound is batch-specific due tovariable water content. Please refer to the vial label or the certificate ofanalysis for the batch-specific molecular weight. The molecular weight providedrepresents the baseline molecular weight without water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic potential of MCC950, a specific inhibitor of NLRP3 inflammasome
Hao Li, et al.
European Journal of Pharmacology, 928, 175091-175091 (2022)
Role of the Nucleotide-Binding Domain-Like Receptor Protein 3 Inflammasome in the Endothelial Dysfunction of Early Sepsis
Luo M, et al.
Inflammation, 43, 1561?1571-1561?1571 (2020)
NLRP3 inflammasome of renal tubular epithelial cells induces kidney injury in acute hemolytic transfusion reactions
Zhixin Liu, et al.
Clinical and Translational Medicine, 11(3), e373-e373 (2021)
Ablation of NLRP3 inflammasome rewires MDSC function and promotes tumor regression
Iosif Papafragkos, et al.
Journal of Separation Science, 13 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service