Skip to Content
Merck
All Photos(1)

Key Documents

O9389

Sigma-Aldrich

Oltipraz

≥98% (HPLC), powder

Synonym(s):

4-Methyl-5-(2-pyrazinyl)-1,2-dithiole-3-thione, BRN 0978110, CCRIS 4048, NSC 347901, RP 35972

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6N2S3
CAS Number:
Molecular Weight:
226.34
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

red

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CC1=C(SSC1=S)c2cnccn2

InChI

1S/C8H6N2S3/c1-5-7(12-13-8(5)11)6-4-9-2-3-10-6/h2-4H,1H3

InChI key

CKNAQFVBEHDJQV-UHFFFAOYSA-N

General description

Oltipraz, a dithiol derivative, is a member of dithiolethiones class of organosulfur compounds. It has anti-tumor, antioxidative, anti- inflammatory and radioprotective properties. It provides protection from numerous carcinogens. Oltipraz functions as a schistosomicide.

Biochem/physiol Actions

Oltipraz is an activator of Nrf2. Nrf2 (NF-E2-related factor 2) is a transcription factor that binds to antioxidant response elements (AREs) and activates these genes. Oltipraz activates Nrf2 and subsequently elevates expression of the detoxification genes encoding anti-oxidant and multidrug resistance-associated proteins to mediate its chemopreventive efficacy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A randomized phase IIb trial of anethole dithiolethione in smokers with bronchial dysplasia
Lam S, et al.
Journal of the National Cancer Institute, 94(13), 1001-1009 (2002)
Alberto Izzotti et al.
Cancer prevention research (Philadelphia, Pa.), 3(1), 62-72 (2010-01-07)
We previously showed that exposure to environmental cigarette smoke (ECS) for 28 days causes extensive downregulation of microRNA expression in the lungs of rats, resulting in the overexpression of multiple genes and proteins. In the present study, we evaluated by
Nrf2 is essential for the chemopreventive efficacy of oltipraz against urinary bladder carcinogenesis
Iida K, et al.
Cancer Research, 64(18), 6424-6431 (2004)
Sang Geon Kim et al.
The Journal of pharmacy and pharmacology, 63(5), 627-635 (2011-04-16)
Oltipraz, a cancer chemopreventive agent, has an anticirrhotic effect in animals. A phase II trial was designed to investigate the preliminary efficacy of oltipraz therapy in liver fibrosis or cirrhosis. Of 83 patients who were randomized to receive placebo, oltipraz
Yoko Yagishita et al.
Antioxidants (Basel, Switzerland), 9(8) (2020-08-14)
The transcription factor NF-E2 p45-related factor 2 (NRF2; encoded by NFE2L2) plays a critical role in the maintenance of cellular redox and metabolic homeostasis, as well as the regulation of inflammation and cellular detoxication pathways. The contribution of the NRF2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service