Skip to Content
Merck
All Photos(7)

Key Documents

F6625

Sigma-Aldrich

Flavin adenine dinucleotide disodium salt hydrate

≥95% (HPLC), powder

Synonym(s):

FAD, FAD-Na2, Riboflavin 5′-adenosine diphosphate disodium salt, FAD-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N9Na2O15P2 · xH2O
CAS Number:
Molecular Weight:
829.51 (anhydrous basis)
Beilstein:
5326842
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

microbial

Quality Level

Assay

≥95% (HPLC)

form

powder

color

yellow to orange-brown

solubility

H2O: soluble-50 mg/mL, clear, slightly orang to deep orange

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)c2cc1C

InChI

1S/C27H33N9O15P2.2Na.H2O/c1-10-3-12-13(4-11(10)2)35(24-18(32-12)25(42)34-27(43)33-24)5-14(37)19(39)15(38)6-48-52(44,45)51-53(46,47)49-7-16-20(40)21(41)26(50-16)36-9-31-17-22(28)29-8-30-23(17)36;;;/h3-4,8-9,14-16,19-21,26,37-41H,5-7H2,1-2H3,(H,44,45)(H,46,47)(H2,28,29,30)(H,34,42,43);;;1H2/q;2*+1;/p-2/t14-,15+,16+,19-,20+,21+,26+;;;/m0.../s1

InChI key

GXTPHHZYFMAGLX-UJXBNFGUSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Flavin adenine dinucleotide disodium salt hydrate (FAD-Na2) is an adenine-containing enzymatic redox cofactor. Also known as flavin cofactors, FAD is critical electron transporter in living systems. They catalyze several 1-2 electron redox reactions. e.g., β-oxidation of fatty acids occurs in the presence of FAD as a cofactor. FAD is one of the two active coenzymes of vitamin B12(riboflavin). FAD displays a significantly shorter excited state lifetime in aqueous solutions than its analog, flavin mononucleotide.

Application

FAD is used to remove reactive oxygen species (ROS) from mammalian cells. The fluorescence mechanism of FAD is used to study energy-dependent intramitochondrial redox potential. FAD is used as a predominant fluorophore to study unstained eosinophils, which exhibit autofluorescence compared to other leucocytes.
Flavin adenine dinucleotide (FAD) is used as a redox cofactor (electron carrier) by flavoproteins including succinate dehydrogenase (complex), α-ketoglutarate dehydrogenase, apoptosis-inducing factor 2 (AIF-M2, AMID), folate/FAD-dependent tRNA methyltransferases, and N-hydroxylating flavoprotein monooxygenases. FAD is a component of the pyruvate dehydrogenase complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruofan Cao et al.
Journal of biomedical optics, 25(1), 1-16 (2020-01-11)
Two-photon fluorescence lifetime imaging microscopy (FLIM) is widely used to capture autofluorescence signals from cellular components to investigate dynamic physiological changes in live cells and tissues. Among these intrinsic fluorophores, nicotinamide adenine dinucleotide (phosphate) (NAD(P)H) and flavin adenine dinucleotide (FAD)-essential
Van Long Nguyen et al.
Analytical biochemistry, 594, 113616-113616 (2020-02-10)
Flavin adenine dinucleotide (FAD) is an active coenzyme of vitamin B2 involved in oxidation and reduction reactions. In this study we have developed a fully automated method for the analysis of FAD by fluorescence detection. FAD is extracted from whole
Virginia Merk et al.
Applied spectroscopy, 75(8), 994-1002 (2021-06-03)
The ultraviolet resonance Raman spectra of the adenine-containing enzymatic redox cofactors nicotinamide adenine dinucleotide and flavin adenine dinucleotide in aqueous solution of physiological concentration are compared with the aim of distinguishing between them and their building block adenine in potential
A N Mayeno et al.
Journal of leukocyte biology, 51(2), 172-175 (1992-02-01)
Unstained human eosinophils exhibit marked autofluorescence in comparison to other leukocytes due to a granule-associated fluorescent substance. Fluorescence spectroscopy of granule extracts reveals excitation maxima at approximately 380 and approximately 450 nm with a single emission at approximately 520, characteristic
Martina Kieninger et al.
Biomolecules, 10(4) (2020-04-15)
Flavin cofactors, like flavin adenine dinucleotide (FAD), are important electron shuttles in living systems. They catalyze a wide range of one- or two-electron redox reactions. Experimental investigations include UV-vis as well as infrared spectroscopy. FAD in aqueous solution exhibits a

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service