O1376
Orotidine 5′-monophosphate trisodium salt
≥99% (HPLC), powder
Synonym(s):
OMP, Orotidylic acid
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C10H10N2O11PNa3
CAS Number:
Molecular Weight:
434.14
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Recommended Products
Quality Level
Assay
≥99% (HPLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
[Na].OC1C(O)C(OC1COP(O)(O)=O)N2C(=O)NC(=O)C=C2C(O)=O
InChI
1S/C10H13N2O11P.Na.H/c13-5-1-3(9(16)17)12(10(18)11-5)8-7(15)6(14)4(23-8)2-22-24(19,20)21;;/h1,4,6-8,14-15H,2H2,(H,16,17)(H,11,13,18)(H2,19,20,21);;
InChI key
PSBNOTIIJINWKE-UHFFFAOYSA-N
Related Categories
Application
Orotidine 5′-monophosphate (OMP) is the direct precursor of uridine 5′-monophosphate (UMP) in pyrimidine biosynthesis. OMP is used to study the specificity, kinetics, structures and mechanisms of orotidine 5′-monophosphate decarboxylase(s) (EC 4.1.1.23).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rumi Ueha et al.
Frontiers in aging neuroscience, 10, 86-86 (2018-04-11)
Background: The incidence of olfactory impairment increases with age; however, the detailed molecular and cellular mechanisms underlying this increase are yet to be determined. Methods: We examined the influence of aging on olfactory receptor neurons (ORNs), which are maintained by
Alexandre Thibodeau et al.
Acta veterinaria Scandinavica, 59(1), 80-80 (2017-12-07)
Campylobacter jejuni is a major cause of foodborne disease having chickens as an important reservoir. Its control at the farm would lower the contamination of the final products and therefore also lower the risk of transmission to humans. At the
Xuebin Tian et al.
BMC microbiology, 20(1), 218-218 (2020-07-22)
The emergence of carbapenem-resistant Klebsiella pneumoniae (CRKP) poses a looming threat to human health. Although there are numerous studies regarding porin alteration in association with the production of ESBLs and/or AmpC β-lactamase, a systematic study on the treatment-emergence of porins
Yang Chen et al.
Physics in medicine and biology, 62(6), 2103-2131 (2017-02-18)
This paper proposes a concise and effective approach termed discriminative feature representation (DFR) for low dose computerized tomography (LDCT) image processing, which is currently a challenging problem in medical imaging field. This DFR method assumes LDCT images as the superposition
Jessica L Rooke et al.
Frontiers in microbiology, 12, 628879-628879 (2021-03-13)
The BAM complex in Escherichia coli is composed of five proteins, BamA-E. BamA and BamD are essential for cell viability and are required for the assembly of β-barrel outer membrane proteins. Consequently, BamA and BamD are indispensable for secretion via
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service