Skip to Content
Merck
All Photos(2)

Key Documents

N7252

Sigma-Aldrich

19-Nortestosterone

Synonym(s):

17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 19-Norandrostenolone, 4-Estren-17β-ol-3-one, Nadrolone, Nandrolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
Beilstein:
2055849
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (TLC)

form

powder

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

chloroform: 50 mg/mL, clear, colorless to light yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1

InChI key

NPAGDVCDWIYMMC-IZPLOLCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

19-Nortestosterone (nandrolone) is an anabolic steriod hormone that acts as an androgen receptor (AR) agonist.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pieter Van Renterghem et al.
Analytica chimica acta, 768, 41-48 (2013-03-12)
Due to their performance enhancing properties, use of anabolic steroids (e.g. testosterone, nandrolone, etc.) is banned in elite sports. Therefore, doping control laboratories accredited by the World Anti-Doping Agency (WADA) screen among others for these prohibited substances in urine. It
J Segura et al.
Forensic science international, 107(1-3), 347-359 (2000-02-26)
Detection of anabolic steroids in hair samples has been possible only in fatal cases or in cases of high-continuous dosages. In order to verify the possibility of detecting an acute administration, a sensitive and specific assay has been developed for
Katrien Venken et al.
Bone, 36(4), 663-670 (2005-03-24)
This study reports the preclinical evaluation of the bone and muscle protective potential of the synthetic androgen 7alpha-methyl-19-nortestosterone (MENTtrade mark), as assessed in the aged orchidectomized rat model. Aged (13-month-old) orchidectomized Wistar rats were treated with different doses of MENT
A Farjam et al.
Analytical chemistry, 63(21), 2481-2487 (1991-11-01)
An immunoaffinity precolumn containing immobilized antibodies raised against the synthetic steroid hormone beta-19-nortestosterone, has been used for the automated sample pretreatment of urine samples containing beta-19-nortestosterone or the related steroids norethindrone and norgestrel. The sample pretreatment system was coupled on-line
R M N Kohler et al.
British journal of sports medicine, 36(5), 325-329 (2002-09-28)
The aim of this review is to analyse the studies on nandrolone metabolism to determine if it is possible for an athlete to test positive for nandrolone without having ingested or injected nandrolone.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service