Skip to Content
Merck
All Photos(3)

Key Documents

N7132

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae

≥95%

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H27N7O14P2Li
CAS Number:
Molecular Weight:
670.37
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

Saccharomyces cerevisiae

Assay

≥95%

form

powder

solubility

H2O: soluble-50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Li].NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5c(N)ncnc45)C(O)C2O

InChI

1S/C21H28N7O14P2.Li.H/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;

InChI key

CUAVZMNXWOTNRC-UHFFFAOYSA-N

Gene Information

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).

Biochem/physiol Actions

Electron acceptor

Other Notes

This is the common form of NAD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard P Ebstein et al.
FEBS letters, 585(11), 1529-1536 (2011-05-12)
Increasing evidence suggests that the nonapeptide, oxytocin (OT), helps shape social and affiliative behaviors not only in lower mammals but also in humans. Recently, an essential mediator of brain OT release has been discovered, ADP-ribosyl cyclase and/or CD38. We have
Michael O Hottiger
FEBS letters, 585(11), 1595-1599 (2011-03-23)
ADP-ribosylation is a covalent post-translational protein modification catalyzed by ADP-ribosyltransferases and is involved in important processes such as cell cycle regulation, DNA damage response, replication or transcription. Histones are ADP-ribosylated by ADP-ribosyltransferase diphtheria toxin-like 1 at specific amino acid residues
Katalin Völgyi et al.
Journal of proteomics, 120, 142-157 (2015-03-19)
The synapse is a particularly important compartment of neurons. To reveal its molecular characteristics we isolated whole brain synaptic (sMito) and non-synaptic mitochondria (nsMito) from the mouse brain with purity validated by electron microscopy and fluorescence activated cell analysis and
Junxiang Yin et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 35(11), 1783-1789 (2015-06-11)
Stroke is one of the leading causes of death. Growing evidence indicates that ketone bodies have beneficial effects in treating stroke, but their underlying mechanism remains unclear. Our previous study showed ketone bodies reduced reactive oxygen species by using NADH
Kazuki Noda et al.
PloS one, 9(11), e110446-e110446 (2014-11-05)
Metabolic disorders, caused by excessive calorie intake and low physical activity, are important cardiovascular risk factors. Rho-kinase, an effector protein of the small GTP-binding protein RhoA, is an important cardiovascular therapeutic target and its activity is increased in patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service