Skip to Content
Merck
All Photos(12)

Key Documents

N7004

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

greener alternative

≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
eCl@ss:
39200202
PubChem Substance ID:
NACRES:
NA.55
Pricing and availability is not currently available.

biological source

yeast

Quality Level

Assay

≥96.5% (HPLC)
≥96.5% (spectrophotometric assay)

form

powder

greener alternative product score

old score: 56
new score: 20
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Use of Renewable Feedstocks
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

application(s)

metabolomics

greener alternative category

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide adenine dinucleotide (NAD) is a vital coenzyme that transcends its traditional role as an "electron carrier," influencing diverse cellular processes. In cell biology, NAD operates as a crucial electron acceptor in the electron transport chain, driving the production of ATP, the cell′s primary energy source. This electron flow sustains basic cellular maintenance and complex signaling pathways. Beyond its energetic role, NAD serves as a dynamic signaling molecule, dictating cellular responses to various stimuli. Fluctuations in NAD concentration trigger events in DNA repair, gene expression, and cell death, allowing cells to adapt to stress and maintain homeostasis.

In biochemical research, NAD emerges as a vital cofactor for enzymes like sirtuins, poly(ADP-ribose) polymerases, and cADP-ribose synthases. These enzymes rely on NAD+ for catalytic activity, influencing processes such as protein deacetylation, DNA repair, and cellular signaling. In metabolomics, NAD plays a key role in metabolic pathways, making it a significant focus for understanding cellular metabolism comprehensively. NAD participates in additional intricate cellular processes, serving as a substrate for ADP-ribosylation—a post-translational modification affecting protein function. It also acts as a precursor for cyclic ADP-ribose, a second messenger molecule involved in calcium mobilization and cell communication. Research on NAD holds promise across various areas, including aging, where its role is actively investigated. Studies also explore its implications in neurodegenerative disorders and metabolic imbalances.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Less Hazardous Chemical Syntheses”, “Safer Solvents and Auxiliaries”, “Enhanced Energy Efficiency”, “Use of Renewable Feedstocks” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Application

β-Nicotinamide adenine dinucleotide hydrate has been used:
  • to prepare NAD+ standard solution for a study on NAD+ metabolomics[1]
  • as a component of MurI assay reaction partial mixture for glutamate racemase reaction[2]
  • as a component of a chromogenic staining solution to stain zebrafish embryos[3]

Biochem/physiol Actions

Electron acceptor

Features and Benefits

Versatile and adaptable for a wide variety of laboratory and research applications.

Packaging

Packaged by solid weight.

Preparation Note

To prepare a solution gentle heating and sonication may be necessary.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
This is the common form of NAD.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana P Gomes et al.
Cell, 155(7), 1624-1638 (2013-12-24)
Ever since eukaryotes subsumed the bacterial ancestor of mitochondria, the nuclear and mitochondrial genomes have had to closely coordinate their activities, as each encode different subunits of the oxidative phosphorylation (OXPHOS) system. Mitochondrial dysfunction is a hallmark of aging, but
Juhyung Lee et al.
Scientific reports, 8(1), 13676-13676 (2018-09-14)
Cyclin-dependent kinase 5 (CDK5) plays a pivotal role in neural development and neurodegeneration. CDK5 activity can be regulated by posttranslational modifications, including phosphorylation and S-nitrosylation. In this study, we demonstrate a novel mechanism by which the acetylation of CDK5 at
Justine Bellier et al.
Cell reports, 30(5), 1400-1416 (2020-02-06)
The use of cetuximab anti-epidermal growth factor receptor (anti-EGFR) antibodies has opened the era of targeted and personalized therapy in colorectal cancer (CRC). Poor response rates have been unequivocally shown in mutant KRAS and are even observed in a majority
Jeerus Sucharitakul et al.
The Journal of biological chemistry, 288(49), 35210-35221 (2013-10-17)
3-Hydroxybenzoate 6-hydroxylase (3HB6H) from Rhodococcus jostii RHA1 is an NADH-specific flavoprotein monooxygenase that catalyzes the para-hydroxylation of 3-hydroxybenzoate (3HB) to form 2,5-dihydroxybenzoate (2,5-DHB). Based on results from stopped-flow spectrophotometry, the reduced enzyme-3HB complex reacts with oxygen to form a C4a-peroxy
Anna Ferretta et al.
Biochimica et biophysica acta, 1842(7), 902-915 (2014-03-04)
Mitochondrial dysfunction and oxidative stress occur in Parkinson's disease (PD), but the molecular mechanisms controlling these events are not completely understood. Peroxisome proliferator-activated receptor-gamma coactivator-1α (PGC-1α) is a transcriptional coactivator known as master regulator of mitochondrial functions and oxidative metabolism.

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service