Skip to Content
Merck
All Photos(1)

Key Documents

N6785

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced disodium salt

~98%, pkg of 10 mg (per vial)

Synonym(s):

β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2
CAS Number:
Molecular Weight:
709.40
Beilstein:
5230241
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

Assay

~98%

Quality Level

form

powder

packaging

pkg of 10 mg (per vial)

solubility

10 mM NaOH: soluble-1 mg/mL, clear, colorless

SMILES string

[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11+,13-,14+,15-,16+,20-,21+;;/m0../s1

InChI key

QRGNQKGQENGQSE-QUWMEQBESA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).

Biochem/physiol Actions

Electron donor

Reconstitution

Solutions should be prepared fresh and used promptly.

Other Notes

Packaged based on NADH content as determined by UV-Vis.
This is the common form of NADH; do not confuse with α-NADH.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arvind Venkat Namuduri et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2269-2286 (2020-01-08)
SUMOylation is a dynamic, reversible, enzymatic drug-targetable post-translational modification (PTM) reaction where the Small Ubiquitin-like Modifier (SUMO) moieties are attached to proteins. This reaction regulates various biological functions like cell growth, differentiation, and it is crucial for maintaining organ homeostasis.
Richard P Ebstein et al.
FEBS letters, 585(11), 1529-1536 (2011-05-12)
Increasing evidence suggests that the nonapeptide, oxytocin (OT), helps shape social and affiliative behaviors not only in lower mammals but also in humans. Recently, an essential mediator of brain OT release has been discovered, ADP-ribosyl cyclase and/or CD38. We have
Weihai Ying
Frontiers in bioscience : a journal and virtual library, 11, 3129-3148 (2006-05-25)
Increasing evidence has indicated that NAD+ and NADH play critical roles not only in energy metabolism, but also in cell death and various cellular functions including regulation of calcium homeostasis and gene expression. It has also been indicated that NAD+
Shin-Ichiro Imai
Biochimica et biophysica acta, 1804(8), 1584-1590 (2009-11-10)
SIR2 (silent information regulator 2) proteins, now called "sirtuins," are an evolutionarily conserved family of NAD-dependent protein deacetylases/ADP-ribosyltransferases. Sirtuins have recently attracted major attention in the field of aging research, and it has been demonstrated that SIR2 and its orthologs
Xiaoling Li et al.
International journal of biological sciences, 7(5), 575-587 (2011-05-27)
Sirtuins are highly conserved NAD+-dependent protein deacetylases and/or ADP-ribosyltransferases that can extend the lifespan of several lower model organisms including yeast, worms and flies. The seven mammalian sirtuins, SIRT1 to SIRT7, have emerged as key metabolic sensors that directly link

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service