N4888
Nutrient Mixture F-12 Ham
With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture
Synonym(s):
Ham’s F-12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Product Name
Nutrient Mixture F-12 Ham, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
NaHCO3: yes
sodium pyruvate: 0.11 g/L
phenol red: yes
L-glutamine: no
HEPES: no
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Nutrient Mixture F-12 is suitable for the growth of primary rat hepatocytes and rat prostate epithelial cells. It is also used in a clonal toxicity assay using Chinese hamster ovary (CHO) cells.
Application
Nutrient Mixture F-12 Ham has been used:
- in Minimum Essential Medium to maintain SH-SY5Y human neuroblastoma cells
- to culture female-originated neuroblastoma cells
- to culture primary retinal pigment epithelium (RPE) cells and RPE cell lines
Reconstitution
Supplement with 0.146 g/L L-glutamine.
also commonly purchased with this product
Product No.
Description
Pricing
related product
Product No.
Description
Pricing
supplement
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yinan Jiang et al.
Molecular immunology, 82, 123-136 (2017-01-11)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL), which is known as a key molecule to induce cancer cell apoptosis, has also been found to participate in the process of ischemia/reperfusion (I/R) injury. Infiltrated macrophages play dual roles in inflammatory injury and
Kambiz Shekdar et al.
Biotechnology letters, 43(5), 949-958 (2021-03-09)
Chromovert® Technology is presented as a new cell engineering technology to detect and purify living cells based on gene expression. The technology utilizes fluorogenic oligonucleotide signaling probes and flow cytometry to detect and isolate individual living cells expressing one or
Roger Oria et al.
Nature, 552(7684), 219-224 (2017-12-07)
Cells can sense the density and distribution of extracellular matrix (ECM) molecules by means of individual integrin proteins and larger, integrin-containing adhesion complexes within the cell membrane. This spatial sensing drives cellular activity in a variety of normal and pathological
Chanan Meydan et al.
Frontiers in immunology, 11, 590870-590870 (2020-11-10)
The COVID-19 pandemic exerts inflammation-related parasympathetic complications and post-infection manifestations with major inter-individual variability. To seek the corresponding transcriptomic origins for the impact of COVID-19 infection and its aftermath consequences, we sought the relevance of long and short non-coding RNAs
Chiara Vidoni et al.
Neurochemistry international, 117, 174-187 (2017-05-24)
Parkinsonian-like motor deficits in Huntington's Disease (HD) patients are associated with abnormal dopamine neurotransmission in the striatum. Dopamine metabolism leads to the formation of oxidized dopamine quinones that exacerbates mitochondrial dysfunction with production of reactive oxygen species (ROS) that eventually
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service