Skip to Content
Merck
All Photos(1)

Documents

I3390

Sigma-Aldrich

IMDM

With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Iscove′s MDM, Iscoves MDM, Iscove’s Modified Dulbecco’s Medium, IMDM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

product name

Iscove′s Modified Dulbecco′s Medium, liquid, sterile-filtered, With sodium bicarbonate, without L-glutamine, suitable for cell culture, suitable for hybridoma

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

endotoxin, tested

components

phenol red: yes
L-glutamine: no
glucose: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Guilbert and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum free medium supplemented with albumin, transferrin, lecithin, and selenium. This medium is a modification of Dulbecco′s Modified Eagle′s Medium (DME) containing selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer, and potassium nitrate instead of ferric nitrate. Further studies demonstrated that Iscove′s Medium would support murine B lymphocytes, hemopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Reconstitution

Supplement with 0.584 g/L L-glutamine

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N N Iscove et al.
The Journal of experimental medicine, 147(3), 923-933 (1978-03-01)
Albumin, transferrin, and lipids can replace serum entirely for support of LPS-stimulated murine B lymphocytes in culture. In the presence of these compounds, growth and maturation to IgM and IgG secretion, induced by lipopolysaccharide (LPS), occurs at the same or
N Vriezen et al.
Biotechnology and bioengineering, 54(3), 272-286 (1997-05-05)
Glutamine is a major source of energy, carbon, and nitrogen for mammalian cells. The amount of glutamine present in commercial mammalian cell media is, however, not necessarily balanced with cell requirements. Therefore, the effects of glutamine limitation on the physiology
Sophie G Kellaway et al.
Haematologica, 106(6), 1569-1580 (2020-04-18)
Hematological malignancies are characterised by a block in differentiation, which in many cases is caused by recurrent mutations affecting the activity of hematopoietic transcription factors. RUNX1-EVI1 is a fusion protein formed by the t(3;21) translocation linking two transcription factors required
Dillon M Voss et al.
Cancer research, 80(21), 4681-4692 (2020-09-16)
Muscleblind-like proteins (MBNL) belong to a family of tissue-specific regulators of RNA metabolism that control premessenger RNA splicing. Inactivation of MBNL causes an adult-to-fetal alternative splicing transition, resulting in the development of myotonic dystrophy. We have previously shown that the
Complete replacement of serum in primary cultures of erythropoietin-dependent red cell precursors (CFU-E) by albumin, transferrin, iron, unsaturated fatty acid, lecithin and cholesterol.
N N Iscove et al.
Experimental cell research, 126(1), 121-126 (1980-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service